Skip to Content
Merck
All Photos(1)

Key Documents

XX42P0060

Millipore

Pellicon® Cassette Standard Acrylic Holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151800
eCl@ss:
32031690
Pricing and availability is not currently available. Contact Technical Service

manufacturer/tradename

Pellicon®

Quality Level

parameter

120 mL min. working volume
4-50 °C temperature
7 bar max. inlet pressure (100 psi)
7 bar max. transmembrane pressure (100 psig)

H

25.4 cm (10 in.)

L

28 cm (11 in.)

W

19 cm (7.6 in.)

weight

11 kg (24 lb)

fitting

3/4 in. inlet/outlet sanitary flange

Related Categories

General description

Pellicon® Cassette Standard Acrylic Holder for Pellicon® 2 Ultrafiltration Cassettes
The Pellicon® Cassette Standard Acrylic Holder is perfect for anyone who needs a benchtop cassette holder versatile enough to support either microporous or ultrafiltration tangential flow filtration.

Application

Concentration, desalting or buffer exchange of:
  • Protein solutions
  • Polysaccharide solutions
  • Virus suspensions


Harvest, washing or clarification of:
  • Cell cultures and lysates
  • Colloidal suspensions
  • Viral cultures

Features and Benefits

  • Leading-edge void- free membranes to match virtually any separation challenge
  • Short flow path for higher flux and higher resolution separation capability
  • Choice of flow channel configuration providing process optimization capability
  • Predictable, fast, scale-up
  • True linear scalability from laboratory size modules to industrial assemblies for processing thousands of liters
  • Brings higher resolution, improved yields and superior back-pressure resistance
  • Ultracel membranes are composite membranes cast on a microporous substrate for defect-free membranes with superior

Benefits
  • Linear Scale-up – a single Pellicon® acrylic holder has up to 10 times the linear scale-up potential in filter area (Note: increased pump capacity is required for scale-up).
  • High Product Recoveries – due to low hold-up volumes.
  • Utilizing Existing Equipment – all equipment is included to connect to your existing peristaltic tubing pump (pump not included). An interchangeable selection of fittings and pumps is also available for changing system capabilities.
  • Production Range – depending upon your needs, between 1 and 500 liter batches can be filtered – in either a matter of minutes or a few hours.

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service