XX1519002
Millipore Base and Cap for Vacuum Filtration
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24
Recommended Products
diam.
90 mm
fitting
6 mm (1/4 in.) outlet tubulated cap sidearm
General description
Millipore Glass Vacuum Filtration Accessory Product
Glass Base and Cap, 90 mm, Glass frit support
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore 90 mm All-Glass Filter Holders Kits.
Glass Base and Cap, 90 mm, Glass frit support
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore 90 mm All-Glass Filter Holders Kits.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service