Skip to Content
Merck
All Photos(1)

Key Documents

TS-10L-BK

Millipore

Bulk Reagent - Rare Specificities & AHG

BIOSCOT Anti-IgG/C3d, Polyspecific Anti-Human Globulin Blood Grouping Reagent

Synonym(s):

Anti-IgG/C3d, PolyspecificAnti-Human Globulin Blood Grouping Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

Note: For further information on price, availability or to place an order, please contact us

biological source

mouse
rabbit

Quality Level

antibody form

unpurified

clone

BRIC-8, polyclonal

form

liquid

packaging

pkg of 10 L

manufacturer/tradename

BIOSCOT

concentration

9-20 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.1-7.3

isotype

IgG
IgM

shipped in

ambient

storage temp.

2-8°C

General description

Colorant: Patent Blue Violet and Tartrazine dyes

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 24 months from date of filtration

Product formulation meets the requirements of Annex IV of the EU IVD directive to support CE registration.

For In Vitro use only

Application

Bulk
For use in Direct and Indirect Anti - Human Globulin tests

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service