Skip to Content
Merck
All Photos(3)

Key Documents

SYMBFEE02

Millipore

Steritest® Symbio Pump Feet

23 mm High feet, Optional accessory for use in laminar flow hood

Synonym(s):

Pump feet

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$87.40

$87.40


Check Cart for Availability

Request more information

Select a Size

Change View
1 EA
$87.40

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

$87.40


Check Cart for Availability

Request more information

material

PVC (Feet)
stainless steel screw(s)

Agency

complies with EP (2.6.1)
complies with JP (4.06)
complies with USP (71)

manufacturer/tradename

Steritest®

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio Pump

shipped in

ambient

General description

The Steritest® Symbio Pump Feet is an optional accessory for the use of Steritest® Symbio pumps in laminar flow hoods. Steritest® Symbio pumps are used for sterility testing in the smallest testing environments, including in a laminar flow hood, biosafety cabinet, cleanroom, or even inside an isolator. The pump feet are made of polyvinyl chloride and used to ease decontamination in the working area.

Application

The Steritest® Symbio Pump Feet is an optional accessory for the use of Steritest® Symbio pumps in laminar flow hoods for streamlining the workflow and increasing safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service