Skip to Content
Merck
All Photos(1)

Key Documents

Z370533

Sigma-Aldrich

Nalgene® bottle-top sterile filter units

filter capacity 500 mL, pore size 0.45 μm, fits 45 mm bottle neck

Synonym(s):

bottletop filtration, rapid-flow filter, sterile culture media filter, vacuum filtration

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
$264.00

$264.00


Available to ship on14 April 2025Details



Select a Size

Change View
12 EA
$264.00

About This Item

UNSPSC Code:
41104914
NACRES:
NB.24

$264.00


Available to ship on14 April 2025Details


material

polyethersulfone membrane
polystyrene housing

sterility

sterile

feature

cap
graduations

packaging

case of 12
pack of 12

manufacturer/tradename

Nalgene® 295-4545

diam.

75 mm

filter capacity

500 mL

funnel capacity

500 mL

neck diam.

45 mm

pore size

0.45 μm

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Nalgene® bottle-top sterile filter units are suitable for the filtration of cell culture media, additives, serum, and buffers.

Features and Benefits

Nalgene® bottle-top sterile filter units have the following benefits:
  • Polyethersulfone (PES) membrane is best suited for tissue culture.
  • Faster results and less waiting time.
  • Non-pyrogenic and endotoxin free.
  • PES exhibits low protein binding, resulting in low chances of loss of protein from media.
  • PES is hydrophilic and thus does not require external wetting agents or surfactants.
  • PES offers higher aqueous flow rates.
  • PES has greater strength, capacity, and chemical resistance than cellulose acetate, nitrocellulose, or nylon.
  • Polystyrene body with graduations.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service