Skip to Content
Merck
All Photos(4)

Key Documents

SYMBBTH01

Millipore

Steritest® Symbio bottle holder (basket and support rod)

for use with Steritest® Symbio pump, this accessory streamlines the workflow and increases safety during sterility testing

Synonym(s):

Bottle mount

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,300.00

$2,300.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EA
$2,300.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

$2,300.00


Check Cart for Availability

Request more information

material

316L stainless steel container (316 L)

Agency

EP (2.6.1)
JP (4.06)
USP (71)

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® Symbio bottle holder is ergonomically positioned around the pump head for easy access in limited-space environments. The bottle holder includes a basket and a support rod. The height and orientation of the bottle holder basket can be adjusted according to the convenience.

Application

Steritest® Symbio bottle holder is an accessory of Steritest® Symbio pumps and used to place the bottle during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service