Skip to Content
Merck
All Photos(2)

Key Documents

SCT240

Sigma-Aldrich

BioTracker Blue TFCH Oxidative Stress Probe

Synonym(s):

Live Cell Imaging Probe

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$614.00

$614.00


Estimated to ship on22 May 2025



Select a Size

Change View
1 VIAL
$614.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.47

$614.00


Estimated to ship on22 May 2025


biological source

human

Quality Level

mol wt

calculated mol wt 280.63 kDa

packaging

vial of 1 mg

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell based assay: suitable
flow cytometry: suitable
immunofluorescence: suitable

detection method

fluorometric

shipped in

dry ice

storage temp.

−20°C

General description

Nephrotoxicity, or drug-induced kidney injury, is a critical consideration when developing new therapeutics. Screening methods currently used to detect and/or assess nephrotoxicity or other cytotoxicity secondary to candidate compounds may not have the molecular specificity to inform results of screening, as traditional toxicity screens are often limited to simple cell viability, morphology, and mitochondrial function readouts. Common screening assays may furthermore lack sensitivity, detecting only severe nephrotoxicity induced by high doses of or long exposures to tested compounds. Mild toxicity in a cell model may present very differently in vivo, necessitating kidney toxicity assays that are sensitive and specific enough to ensure therapeutic safety.One approach to enhancing the utility of toxicity screening assays is to measure oxidative stress, which is often more sensitive to mild drug injury. While certain indicators of oxidative stress such as increases in reactive oxygen species (ROS) or depletion of reductants are transient, one durable change to kidney (and other) cells is the post-injury carbonylation of cellular biomolecules. 4-trifluoromethyl-7-hydrazinyl-2H-chromen-2-one (TFCH) is intended to enhance sensitivity to cell injury induced by nephrotoxic or cytotoxic substances in renal system applications including in vivo and in vitro models such as Madin-Darby Canine Kidney (MDCK) cells.Spectral Properties Fluorescence images were obtained by λex = 405 nm and emission at 550 to 650 nm.ReferenceMukherjee, et al. "A Novel Fluorogenic Assay for the Detection of Nephrotoxin-induced Oxidative Stress in Live Cells and Renal Tissue." ACS Sensors 6 (2021): 2523-2528.

Application

Purity: > 98% confirmed by HPLC. Identification confirmed by HNMR, LC-MS and elemental analysis.

Features and Benefits

Fluorescent live cell probe for visualizing oxidative stress, cytotoxicity, nephrotoxicity from candidate compounds and other substances.

Physical form

Lyophilized. Yellow solid.

Storage and Stability

Store at -20°C, desiccated and protected from light.Note: Centrifuge vial briefly to collect contents at bottom of vial before opening

Other Notes

Live cell fluorescent imaging

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service