Skip to Content
Merck
All Photos(3)

Key Documents

S7450-M

Sigma-Aldrich

OxyIHC Oxidative Stress Detection Kit

The OxyIHC Oxidative Stress Detection Kit contains the chemical & immunological reagents necessary to detect protein oxidation in various tissues from a variety of organs & animal species.

Synonym(s):

Oxidative Stress Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

50 REACTIONS
$1,140.00

$1,140.00


Estimated to ship on29 May 2025



Select a Size

Change View
50 REACTIONS
$1,140.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

$1,140.00


Estimated to ship on29 May 2025


Quality Level

manufacturer/tradename

OxyIHC

technique(s)

immunohistochemistry: suitable

detection method

colorimetric

General description

Reactive oxygen species (ROS) have been implicated in pathological processes including cancer,
apoptosis, aging, neurodegenerative diseases, chronic inflammatory diseases, pulmonary diseases, and
cardiovascular diseases (for reviews, see ref. 1-4). Living organisms are continually exposed to potentially
harmful oxygen free radicals that are generated thru normal cellular functions as well as from environmental
factors (1, 5-7). Yet, these reactive molecules are mitigated by the presence of antioxidants and therefore
organisms exist with a delicate balance between oxidants and antioxidants (7-9). During times of oxidative
stress there is an imbalance in favor of the oxidants. The end result of which is that free radicals are able to
attack and modify sub-cellular components including nucleic acids, lipids and proteins (7, 10-12).
Proteins are one of the major targets of reactive species. Oxidation of proteins modifies the side chains of
methionine, histidine, and tyrosine and forms cysteine disulfide bonds (16-19). Metal catalyzed oxidation of
proteins introduces carbonyl groups (aldehydes and ketones) at lysine, arginine, proline or threonine
residues in a site-specific manner (16, 20-22). The oxidative modification of proteins can modulate
biochemical characteristics of proteins such as enzymatic activity (21-23), DNA binding activities of
transcription factors (24-26), and the susceptibility to proteolytic degradation (12, 25-28). While a
relationship between protein oxidation and aging has been suggested (29-31), little is known about the
importance of oxidative modification of individual proteins in the pathophysiology of free radical mediated
processes. Not surprisingly, carbonyl modification of proteins has become a key biomarker for the
identification of oxidative stress.
The OxyIHC Oxidative Stress Detection Kit (S7450) contains the chemical and immunological reagents
necessary to detect protein oxidation in various tissues from a variety of organs and animal species. The
OxyIHC test method involves chemical derivatization of protein carbonyl groups with 2,4-
dinitrophenylhydrazine (DNPH). This chemical reaction results in proteins being covalently coupled to DNP
at their carbonyl sites. The DNP-derivatized proteins are then detected using an antibody that specifically
binds to the DNP moiety. Subsequent incubation with biotin conjugated secondary antibody, streptavidin
conjugated HRP, and development using a 3,3’ diaminobenzidine (DAB) staining allows
immunohistochemical detection of protein oxidation.

Application

Research Category
Apoptosis & Cancer

Neuroscience
Research Sub Category
Neurodegenerative Diseases

Aging
The OxyIHC Oxidative Stress Detection Kit contains the chemical & immunological reagents necessary to detect protein oxidation in various tissues from a variety of organs & animal species.

Components

10X Wash Buffer, One 25 mL bottle

10X Antigen Retrieval Buffer, One 1.25 mL bottle

5X Blocking Buffer, One 2.5 mL bottle

Primary Antibody, One 125 mL vial

DAB-A, One 500 mL vial

DAB-B, One 12 mL bottle

Antibody Diluent, One ready to use 12.5 mL bottle

Biotinylated Secondary Antibody, One ready to use 12.5 mL bottle

Streptavidin conjugated HRP, One ready to use 12.5 mL bottle

Hematoxylin, One ready to use 12.5 mL bottle

DNPH Solution, One ready to use 12.5 mL bottle

Derivatization Control Solution, One ready to use 12.5 mL bottle

Storage and Stability

The OxyIHC kit components should be stored at 2-8ºC. Kit components are stable for 6 months from the
date of receipt if stored and handled properly.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Met. Corr. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. what antigen retrieval does the kit provide?

    1 answer
    1. This kit ships with an antigen retrieval buffer. The protocol is listed below and is also included in the user guide.
      Please see the following link to access the user guide:
      https://www.emdmillipore.com/product/OxyIHC-Oxidative-Stress-Detection-Kit,MM_NF-S7450#documentation

      Please see the Antigen Retrieval steps below:
      1. Obtain slides with deparaffinized and rehydrated tissue (as indicated in the User Guide).
      2. Perform antigen unmasking of tissue by adding 250 µL of 1X Antigen Retrieval Buffer per slide.
      3. Place a glass coverslip on top of the slide and incubate in a steamer for 20 minutes.
      4. Allow the slide to cool down for 15 minutes at room temperature.
      5. Wash the tissue by incubating it in 250 µL of 1X Wash Buffer for 5 minutes. Repeat the wash 2
      additional times

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service