SCHVU02RE
Stericup-HV Sterile Vacuum Filtration System
250 mL process volume, 0.45 µm pore size, PVDF membrane, pkg of 12, radio-sterilized
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.22
Recommended Products
material
PVDF membrane
polystyrene
Quality Level
sterility
sterile
feature
holdup volume 2.4 mL
hydrophilic
manufacturer/tradename
Stericup®
parameter
250 mL process volume
45 °C max. temp.
technique(s)
sterile filtration: suitable
H
164 mm
diam.
12.5 cm
filter diam.
73 mm
filtration area
40 cm2
funnel capacity
250 mL
matrix
Durapore®
pore size
0.45 μm nominal pore size
0.45 μm pore size
test parameters
sample volume: 250 mL
fitting
inlet funnel
outlet flask (Receiver)
shipped in
ambient
General description
250 mL bottle with cap
The Stericup® vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop filter unit with a receiver flask for processing and storing volumes from 150 mL to 1,000 mL in a single, ready-to-use format. Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device. The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop bottle-top devices with your own sterile capture flask or bottle.
Features & Benefits
Millipore Express PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications
Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component
Features & Benefits
Millipore Express PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs
Durapore membrane for ultra-low protein binding for minimum absorption and protein loss
Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
Stable base and easy grip design of the Stericup device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
Applications
Sterilization of Aqueous Solutions (150 mL to 1 L); Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component
Application
250 mL process volume, 0.45 µm pore size, PVDF membrane, pkg of 12, radio-sterilized
Principle
Vacuum
Physical form
Color Code: Clear with blue adapter
Preparation Note
Sterilization Method
Gamma irradiation
Gamma irradiation
Legal Information
Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service