Skip to Content
Merck
All Photos(1)

Key Documents

S7113

Sigma-Aldrich

DAPI/Antifade Solution, Ready to Use

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$331.00

$331.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$331.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

$331.00


Estimated to ship on29 May 2025


Request a Bulk Order

manufacturer/tradename

Chemicon®

Quality Level

detection method

fluorometric

shipped in

dry ice

General description

DAPI: 4′, 6-Diamino-2-phenylindole dihydrochloride. Antifade Solution: p-Phenylenediamine and glycerol in PBS solution. 0.4μg/mL DAPI suspended in Antifade. The product is ready to add to fixed slides or cells cultures.

Application

A nuclear fluorescent stain used for counterstaining paraffin-embedded, formalin-fixed tissue, metaphase chromosomes, or interphase nuclei that have been hybridized with probes detected with Fluorescein, or equivalent stains. Antifade prevents the permanent loss of fluorescence or fluorescent signal quenching due to photobleaching and prolonged exposure to high intensity light sources.

DAPI binds the minor groove of DNA at AT clusters and binds preferentially to ds DNA (Biochemistry 26:4545, 1987).

Materials Provided

Each vial contains 1 ml of DAPI/Antifade Solution

Warnings and Precautions

DAPI is a potential carcinogen. Antifade is a potential toxin and irritant. Use extreme caution. Avoid contact with eyes, skin and mucous membranes. Use extreme caution. Wear gloves and a lab coat when handling. Follow your institution′s guidelines for hazardous waste disposal. Keep tightly capped and limit exposure to light.



Related Products

Antifade Solution S7114

Propidium Iodide Solution S7109

Propidium Iodide/Antifade Solution S7112

Physical form

Liquid PBS pH 8.0 containing ~50% glycerol and 0.4μg/mL of DAPI and 2-7mM phenylenediamine.

Storage and Stability

Store at -20ºC in undiluted aliquots for up to 1 year from date of receipt. Protect from light

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Creation of Biofunctionalized Micropatterns on Poly(methyl methacrylate) by Single-Step Phase Separation Method.
Ho QP, Wang SL, Wang MJ
ACS Applied Materials & Interfaces null
Observation of chondrocyte aggregate formation and internal structure on micropatterned fibroin-coated surface.
Naoyoshi D Kachi,Akihisa Otaka,Seungwoo Sim,Yoshihiko Kuwana,Yasushi Tamada,Junko Sunaga et al.
Bio-medical materials and engineering null
Yusuke Kambe et al.
Biomaterials, 31(29), 7503-7511 (2010-07-21)
Initial chondrocyte-silk fibroin interactions are implicated in chondrogenesis when using fibroin as a scaffold for chondrocytes. Here, we focused on integrin-mediated cell-scaffold adhesion and prepared cell adhesive fibroin in which a tandem repeat of the Arg-Gly-Asp-Ser (RGDS) sequence was genetically
Mohammadhossein Khorraminejad-Shirazi et al.
Stem cell research & therapy, 11(1), 45-45 (2020-02-06)
Mesenchymal stromal cell (MSC) stemness capacity diminishes over prolonged in vitro culture, which negatively affects their application in regenerative medicine. To slow down the senescence of MSCs, here, we have evaluated the in vitro effects of 5-aminoimidazole-4-carboxamide ribonucleotide (AICAR), an
Kuan-Jung Chen et al.
Journal of cell science, 133(3) (2020-01-15)
Cell migration requires the coordination of multiple signaling pathways involved in membrane dynamics and cytoskeletal rearrangement. The Arf-like small GTPase Arl4A has been shown to modulate actin cytoskeleton remodeling. However, evidence of the function of Arl4A in cell migration is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service