Skip to Content
Merck
All Photos(2)

Key Documents

QGARDT2X1

Milli-Q

Q-Gard® Purification Cartridge

Q-Gard® Purification Cartridge

For Milli-Q® Advantage A10 / Reference systems connected to DI water feed

Synonym(s):

Q-Gard® T2 Purification Cartridge

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$941.00

$941.00


Available to ship on09 April 2025Details


Request more information

Select a Size

Change View
1 EA
$941.00

About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

$941.00


Available to ship on09 April 2025Details


Request more information

Quality Level

feature

can produce high resistivity water from purification media

packaging

pkg of 1 unit

manufacturer/tradename

Q-Gard®

resistivity

>18.0 MΩ-cm (at 25 °C)

capacity

≤53 g ion exchange capacity (NaCl)(Dynamic Capcity is greater than 34 g NaCl)

compatibility

for use with Milli-Q® Advantage A10
for use with Milli-Q® Reference

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Q-Gard® T2 purification cartridge is a replacement cartridge designed to remove ionic and organic contaminants down to trace levels from DI water. The cartridge is constructed with clean and high-purity materials to minimize leaching of extractable contaminants. It contains a microfilter for particulates removal, activated carbon for organics adsorption, and mixed-bed ion-exchange resin for ion removal. Q-Gard® T2 Purification Cartridge can be used with Milli-Q® Advantage A10 / Reference water purification systems.

Application

Removal of ionic and organic water contaminants.

Features and Benefits

  • Genuine consumable for the replacement of Q-Gard® QGARDT2X1 purification cartridge.
  • Generates ionic and organic contaminant-free water by ion exchange, activated carbon, and depth filtration technologies.
  • Certificate of Quality available online.

Components

Virgin ion-exchange resins for deionization

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-GARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–10 of 10 Questions  
  1. What is the proper disposal method for Water filter products at the end of their life? In the past, there was a recycling program, but since that system has ended, what is the appropriate approach for disposal?

    1 answer
    1. The recycling program was discontinued due to low usage. It closed because of low activity and infrequent consumable returns. The materials being non-hazardous can be disposed of with standard laboratory waste.

      Helpful?

  2. What are the storage conditions for this product?

    1 answer
    1. Recommended storage conditions: temperatures from 5 to 30°C; humidity between 10 and 80%. The original sealed bag (if provided) must be kept intact before the first use to protect the purification element from foreign matter.

      Helpful?

  3. How are consumables changed?

    1 answer
    1. To help you to change your system's consumables, several video tutorials are available on our YouTube channel: https://youtube.com/playlist?list=PL2TUriaAaEA_IXDCnPbDk6OmrwptjwOHD. For high-flow systems, please keep in mind that all consumable changes must be approved by a 3-digit code that only the person in charge of the system knows.

      Helpful?

  4. How can I know which consumables can be used with my system?

    1 answer
    1. To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.

      Helpful?

  5. What steps are you taking towards improved sustainability?

    1 answer
    1. Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.

      Helpful?

  6. Can I arrange to have my consumables automatically ordered and delivered once a year?

    1 answer
    1. It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).

      Helpful?

  7. Can consumables be opened and refilled?

    1 answer
    1. Unfortunately, this is not possible because of safety issues and the need to guarantee the quality of consumables and their interior purification components. Our consumables are manufactured in an ISO 9001-certified facility France in agreement with high quality standards.

      Helpful?

  8. How frequently do the various consumables need to be changed?

    1 answer
    1. The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.

      Helpful?

  9. Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?

    1 answer
    1. Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.

      Helpful?

  10. What is the consumables delivery time?

    1 answer
    1. Refer to the consumable's Product page for a first estimation of its expected delivery time and contact customer service for more information.

      Helpful?

Reviews

Active Filters

  1. Kansas
    • Reviews 3
    • Votes 2
    5 out of 5 stars.

    For Adavantage A10 system

    Good filter for our water system. Just wish Millipore had a usage amount for the filters. They only state the filters are good for one year. (10,000L?, 100,000L?, more/less?). Our SOP states to change the filters every 6 months. I need a volume to justify to change to one year. Anyway, the filters after about 40,000L still maintain integrity.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service