Skip to Content
Merck
All Photos(1)

Key Documents

PX0995

Supelco

o-Phosphoric Acid

85% W/W, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Phosphoric acid, Orthophosphoric Acid, Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
MDL number:
UNSPSC Code:
12352301
Pricing and availability is not currently available.

Product Name

o-Phosphoric Acid 85% W/W, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

grade

ACS reagent

Quality Level

Agency

meets USP/NF

vapor density

3.4 (vs air)

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

form

liquid

concentration

85% phosphoric acid

impurities

≤0.001% Heavy metals (as Pb)
≤0.001% Insoluble matter
≤0.001% Volatile acids (as acetic acid)
≤0.002% Antimony (Sb)
≤0.002% Calcium (Ca)
≤0.002% Magnesium (Mg)
≤0.003% Iron (Fe)
≤0.003% Sulfate (SO4)
≤0.005% Potassium (K)
≤0.025% Sodium (Na)
≤0.5 ppm Manganese (Mn)
≤1 ppm Arsenic (As)
≤3 ppm Chloride (Cl)
≤5 ppm Nitrate (NO3)

color

APHA: ≤10

pH

<0.5 (20 °C, 100 g/L in H2O)

bp

158 °C (lit.)

mp

~40 °C (lit.)

density

1.685 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
PX0995P-5 and PX0995P-1 are packaged in poly coated glass bottles
Click here for printer friendly instructions for adapting a hand pump to the 20L HDPE drum.

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

Analysis Note

Antimony (Sb): 0.002% max
Arsenic (As): 1ppm max
Assay: 85.0% min
Calcium (Ca): 0.002% max
Chloride (Cl): 3ppm max
Color (APHA): 10 max
Form: Clear liquid
Heavy metals (as Pb): 0.001% max
Insoluble matter: 0.001% max
Iron (Fe): 0.003% max
Magnesium (Mg): 0.002% max
Manganese (Mn): 0.5ppm max
Nitrate (NO3): 5ppm max
Potassium (K): 0.005% max
Reducing substances: To pass test
Sodium (Na): 0.025% max
Sulfate (SO4): 0.003% max
Volatile acids (as acetic acid): 0.001% max

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service