Skip to Content
Merck
All Photos(2)

Documents

1.00565

Supelco

ortho-Phosphoric acid

99%, cryst. for analysis EMSURE®

Synonym(s):

Phosphoric acid, Orthophosphoric acid, o-Phosphoric acid, Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-633-2
NACRES:
NA.21

product name

ortho-Phosphoric acid 99%, cryst. for analysis EMSURE®

vapor density

3.4 (vs air)

Quality Level

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

product line

EMSURE®

Assay

≥99% (acidimetric)

form

solid

potency

2740 mg/kg LD50, skin (Rabbit)

concentration

85% phosphoric acid

impurities

≤20 ppm Substances reducing potassium permanganate (as O)
≤20 ppm Volatile acids (as CH3COOH)

pH

<0.5 (20 °C, 100 g/L in H2O)

bp

158 °C (lit.)

mp

~40 °C (lit.)

solubility

750-850 g/L

density

1.685 g/mL at 25 °C (lit.)
1.84 g/cm3 at 38 °C

anion traces

chloride (Cl-): ≤2 ppm
fluoride (F-): ≤0.5 ppm
nitrate (NO3-): ≤2 ppm
phosphite, hypophosphite (as H3PO3): ≤500 ppm

cation traces

As: ≤1 ppm
Ca: ≤20 ppm
Cd: ≤1 ppm
Co: ≤1 ppm
Cu: ≤1 ppm
Fe: ≤10 ppm
K: ≤5 ppm
Mg: ≤1 ppm
Mn: ≤1 ppm
Na: ≤50 ppm
Ni: ≤1 ppm
Pb: ≤1 ppm
Sb: ≤1 ppm
Zn: ≤1 ppm

storage temp.

2-30°C

SMILES string

OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

Analysis Note

Assay (acidimetric): ≥ 99 %
Identity: passes test
Chloride (Cl): ≤ 2 ppm
Nitrate (NO₃): ≤ 2 ppm
Phosphite and Hypophosphite (as H₃PO₃): ≤ 500 ppm
Fluoride (F): ≤ 0.5 ppm
As (Arsenic): ≤ 1 ppm
Ca (Calcium): ≤ 20 ppm
Cd (Cadmium): ≤ 1 ppm
Co (Cobalt): ≤ 1 ppm
Cu (Copper): ≤ 1 ppm
Fe (Iron): ≤ 10 ppm
K (Potassium): ≤ 5 ppm
Mg (Magnesium): ≤ 1 ppm
Mn (Manganese): ≤ 1 ppm
Na (Sodium): ≤ 50 ppm
Ni (Nickel): ≤ 1 ppm
Pb (Lead): ≤ 1 ppm
Sb (Antimony): ≤ 1 ppm
Zn (Zinc): ≤ 1 ppm
Volatile acids (as CH₃COOH): ≤ 20 ppm
Substances reducing potassium permanganate (as O): ≤ 20 ppm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service