Skip to Content
Merck
All Photos(2)

Key Documents

PR0GTXL0S2

Milli-Q

Progard® XL Pretreatment Pack

For optimal protection of the reverse osmosis (RO) membrane from tap water components.

Synonym(s):

Progard® XL-S Pretreatment Pack

Sign Into View Organizational & Contract Pricing

Select a Size

2 PKG
$2,160.00

$2,160.00


Available to ship on11 April 2025Details


Request more information

Select a Size

Change View
2 PKG
$2,160.00

About This Item

UNSPSC Code:
41104212
NACRES:
NB.85

$2,160.00


Available to ship on11 April 2025Details


Request more information

material

carbon cartridge (activated )
cartridge (Polyphosphate for anti-scaling protection)

Quality Level

packaging

pkg of 2 units

manufacturer/tradename

Progard®

parameter

2-40 °C temp. range
4.2 L/min flow rate
6 bar max. pressure

H × W × D

20.8 in. × 6.1 in. × 3.1 in.

fitting

11 mm female inlet and outlet (O-rings)

compatibility

for use with AFS®
for use with Elix® Large
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
for use with RiOs Large

Looking for similar products? Visit Product Comparison Guide

General description

The Progard® XL-S pretreatment pack pretreats feed water by removing large particles, organic compounds and free chlorine from tap water. It protects the reverse osmosis (RO) membrane against mineral scaling, organic fouling, and chlorine oxidation.
The pretreatment pack includes prefiltration to remove particles, activated carbon to remove chlorine, and an anti-scaling agent to minimize the risk of scale formation.
For use with larger RiOs, Elix® and AFS® E water purification systems.

Application

The packs protect and maintain good performance of the reverse osmosis membrane.

Features and Benefits

  • Polyphosphate provides anti-scaling protection.
  • Silver-impregnated activated carbon reduces free chlorine and provides bacteriostatic protection.
  • Depth-filtration removes particles and colloids.
  • e-Sure tag RFID technology for instant visibility on pack parameters, automatic pack parameters archiving and full traceability.

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–9 of 9 Questions  
  1. What are the storage conditions for this product?

    1 answer
    1. Recommended storage conditions: temperatures from 5 to 30°C; humidity between 10 and 80%. The original sealed bag (if provided) must be kept intact before the first use to protect the purification element from foreign matter.

      Helpful?

  2. How are consumables changed?

    1 answer
    1. To help you to change your system's consumables, several video tutorials are available on our YouTube channel: https://youtube.com/playlist?list=PL2TUriaAaEA_IXDCnPbDk6OmrwptjwOHD. For high-flow systems, please keep in mind that all consumable changes must be approved by a 3-digit code that only the person in charge of the system knows.

      Helpful?

  3. How can I know which consumables can be used with my system?

    1 answer
    1. To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.

      Helpful?

  4. What steps are you taking towards improved sustainability?

    1 answer
    1. Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.

      Helpful?

  5. Can I arrange to have my consumables automatically ordered and delivered once a year?

    1 answer
    1. It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).

      Helpful?

  6. Can consumables be opened and refilled?

    1 answer
    1. Unfortunately, this is not possible because of safety issues and the need to guarantee the quality of consumables and their interior purification components. Our consumables are manufactured in an ISO 9001-certified facility France in agreement with high quality standards.

      Helpful?

  7. Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?

    1 answer
    1. Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.

      Helpful?

  8. What is the consumables delivery time?

    1 answer
    1. Refer to the consumable's Product page for a first estimation of its expected delivery time and contact customer service for more information.

      Helpful?

  9. How frequently do the various consumables need to be changed?

    1 answer
    1. The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service