Skip to Content
Merck
All Photos(1)

Key Documents

PICT

Sigma-Aldrich

Protease Inhibitor Cocktail Tablets

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$305.00

$305.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$305.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NC.07
solubility:
water: soluble

$305.00


Estimated to ship on23 May 2025


Request a Bulk Order

solubility

water: soluble

Quality Level

shipped in

wet ice

Application

All

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service