Key Documents
P0SIC0NT1
Millipore
Milliflex® Positive Control Tool
To verify Milliflex® Rapid System Reagent Spray and Enumeration Functionality
About This Item
UNSPSC Code:
41116500
NACRES:
NA.85
Pricing and availability is not currently available. Contact Technical Service
material
aluminum
packaging
pkg of 1 kit
manufacturer/tradename
Milliflex®
technique(s)
Rapid Sterility Testing: suitable
application(s)
microbiology
pharmaceutical
sterility testing
compatibility
for use with Milliflex® Rapid
General description
Milliflex® Positive Control Tool is designed to verify Milliflex® Rapid System reagent spray and microbial enumeration functionality.
Milliflex® Rapid systems are used to detect live microorganism in the tested sample. The positive control will help determine the efficiency of the reagent spray onto the sample and the corresponding microbial enumeration.
Application
Milliflex® Positive Control Tool is used to verify Milliflex® Rapid System reagent spray and microbial enumeration functionality.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Articles
Milliflex® Rapid System 2.0 accelerates microbial detection in quality control labs.