Skip to Content
Merck
All Photos(1)

Key Documents

MISP00015

Millipore

Microfil® Syringe Spare Part Kit

For use with Microfil® filtration systems

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$169.00

$169.00


Estimated to ship on31 May 2025


Request more information

Select a Size

Change View
1 EA
$169.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

$169.00


Estimated to ship on31 May 2025


Request more information

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Microfil®

input

sample type food(s)
sample type beverage(s)

application(s)

bioburden testing
clinical
environmental
food and beverages
industrial qc
water monitoring

compatibility

for use with Microfil® filtration systems

shipped in

ambient

Related Categories

General description

The syringe spare part kit is an additional spare part of the Microfil® filtration system. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. The syringe spare part kit aids as a support system for the efficient working of the filtration system.

Application

The syringe spare part kit is an additional accessory of the Microfil® filtration system used in general microbiological analysis of beer, bottled water, pharmaceutical products analysis, environmental analysis, bioburden testing, and water monitoring.

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service