Skip to Content
Merck

MCR4001FS1

Millipore

Millistak+® Pod Depth Filter

CR40, filtration area 0.11 m², Process Scale

Synonym(s):

Millistak+ Pod Depth Filter, CR40 media series, 0.11 m 2 surface area, 1/pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690
Pricing and availability is not currently available. Contact Technical Service

Product Name

Millistak+® Pod Depth Filter, CR40, cellulose, filtration area 0.11 m2, hydrophilic

material

carbon (active)
cellulose
polypropylene

Quality Level

description

Activated carbon is a very versatile process aid and has a high affinity and adsorption capacity for odorous compounds and color bodies over a wide range of molecular weights

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

1 bar max. differential pressure (15 psid) at 80 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Reverse)

H

32 cm (12.5 in.)

L

62 cm (24.2 in.)

filtration area

0.11 m2

impurities

<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)

total organic carbon (TOC) residue

1300-1700 ppb

input

sample type liquid

conductivity

4.36-4.57 μS/cm at 25 °C (product water)

filter grade

CR40

General description

Device Configuration: multilayer Pod

Application

Activated carbon is a very versatile process aid and has a high affinity and adsorption capacity for odorous compounds and color bodies over a wide range of molecular weights

Specifications

Filter Grade: CR40

Preparation Note

Sterilization Method
Integrity is maintained after 1 autoclave cycle of 60 minutes @ 123 °C

Analysis Note

Product Water Conductivity at 25°C: post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 liters per ft2 (100 L/m2) of media surface area
Total Organic Carbon (TOC) Residue: post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 liters per ft2 (100 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service