Skip to Content
Merck
All Photos(1)

Key Documents

MAWP037A0

Millipore

Aerosol Monitor, 37 mm, mixed cellulose esters, 0.8 µm, white, with pad

Microscopic particle analysis and sample collection

Synonym(s):

Air Quality Monitoring Device

Sign Into View Organizational & Contract Pricing

Select a Size

50 PKG
$269.00

$269.00


Check Cart for Availability


Select a Size

Change View
50 PKG
$269.00

About This Item

UNSPSC Code:
41116116
eCl@ss:
32161002
NACRES:
NB.24

$269.00


Check Cart for Availability

material

cellulose membrane
mixed cellulose esters (MCE) membrane
plain filter
polystyrene
white filter

Quality Level

sterility

non-sterile

filter diam.

37 mm

filtration area

9 cm2

pore size

0.8 μm pore size

fitting

female Luer inlet slip
female Luer outlet slip

application(s)

environmental

shipped in

ambient

General description

Filter Code: AAWP
Millipore′s 37 mm Aerosol Analysis Monitors are transparent, disposable, plastic filter holders pre-assembled with Type MF (mixed cellulose esters) Millipore filters. A thin cellulose support seals the filter between monitor sections to distribute sample flow evenly over the filter surface. The three-part design holds the filter in-place when the top section is removed for open aerosol sampling.

The Average Background particle Count (ABC) value is marked on each monitor package.

- Pre-assembled with filters in-place
- Available with matched-weight membranes

Applications:
Aerosol analysis monitors are available with 0.45 µm and 0.8 µm cellulose esters membranes. Matched weight monitors contain two filters matched in weight to within 0.1 mg, eliminating test filter pre-weighing in gravimetric analysis.

Application

Product Filters Out: Asbestos, trace metals, lead, lead compounds, copper, particulate gases, sulfur dioxide, arsenic

Principle

Vacuum

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service