Skip to Content
Merck
All Photos(2)

Documents

M00003700

Millipore

Empty Monitor Case without ring

Close monitor inlet and outlet

Synonym(s):

Monitor Case

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
eCl@ss:
32161002
NACRES:
NB.24

material

polystyrene

Quality Level

sterility

non-sterile

filter diam.

37 mm

fitting

female Luer inlet slip
female Luer outlet slip

application(s)

environmental

shipped in

ambient

General description

Aerosol Monitor Cases - M00003700, M000037A0



•Empty aerosol analysis monitors

•For use with Type MF (mixed cellulose esters) Millipore filters



Millipore′s 37 mm Aerosol Analysis Monitors are transparent, disposable, plastic filter holders for use with Type MF (mixed cellulose esters) Millipore filters. A thin cellulose support seals the filter between monitor sections to distribute sample flow evenly over the filter surface.



A three-part design holds the filter in-place when the top section is removed for open aerosol sampling. The Average Background particle Count (ABC) value is marked on each monitor package.



Applications:

Aerosol analysis monitors are available with 0.45 µm and 0.8 µm cellulose esters membranes. Matched weight monitors contain two filters matched in weight to within 0.1 mg, eliminating test filter pre-weighing in gravimetric analysis.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service