Skip to Content
Merck
All Photos(1)

Key Documents

MAHAN4550

Millipore

MULTISCREEN®-HA MCE 0.45um Clear Non-sterile 50/pk

Sign Into View Organizational & Contract Pricing

Select a Size

MAHAN4550
$1,950.00

$1,950.00


Estimated to ship on12 May 2025



Select a Size

Change View
MAHAN4550
$1,950.00

About This Item

UNSPSC Code:
12352200
NACRES:
NB.15

$1,950.00


Estimated to ship on12 May 2025


Quality Level

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service