Skip to Content
Merck
All Photos(1)

Key Documents

MABT394

Sigma-Aldrich

Anti-Plakophilin-2 Antibody, clone 8H6

clone 8H6, from mouse

Synonym(s):

Plakophilin-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$778.00

$778.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$778.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$778.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8H6, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PKP2(5318)

General description

Plakophilin-2 (PKP2) is a member of the Armadillo/β-catenin family. It contains eight ARM repeats and can exist in two slice isoforms (2a and 2b). Plakophilin-2 is highly abundant in simple and stratified epithelial cells; but is also expressed in non-epithelial cells, such as the myocardium and lymph node follicles. In epithelial cells, it is localized to desmosomal plaques and plays an important role in cell-cell adhesion processes. Plakophilin-2 also maintains the cell-cell junctions of cardiac tissue, and is essential for proper cardiac function. Mutations in the plakophilin-2 gene cause arrhythmogenic right ventricular cardiomyopathy 9, a condition characterized by partial breakdown of the myocardium of the right ventricle, electrical instability, and sudden death. Plakophilin-2 has also been implicated in the development of cancer.

Immunogen

MBP-tagged recombinant protein corresponding to human Plakophilin-2.

Application

Anti-Plakophilin-2 Antibody, clone 8H6 detects level of Plakophilin-2 & has been published & validated for use in Western Blotting, ICC.
Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in A431 and UM-SCC-38 cells. (Sobolik-Delmaire, T., et al. (2010). Journal of Investigative Dermatology. 130:2638–2646.)
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Plakophilin-2 in 10 µg of A431 cell lysate.

Target description

~97 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service