Skip to Content
Merck
All Photos(3)

Key Documents

ABT283

Sigma-Aldrich

Anti-Cadherin-11/OB Cadherin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cadherin-11, OSF-4, Osteoblast cadherin, OB-cadherin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$508.00

$508.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$508.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$508.00


Estimated to ship on27 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CDH11(1009)

General description

Cadherins constitute a family of transmembrane glycoproteins involved in Ca2+-dependent cell-cell interactions. The members of this family are differentially expressed in various tissues. They function in the maintenance of tissue integrity and morphogenesis. The cadherins generally contain five extracellular repeats, a transmembrane domain and a cytoplasmic tail that binds to the catenin family of cytoskeletal anchoring proteins which also function as signal transducers. The extracellular domains are responsible for the specificity of homophilic interactions between cells expressing the same cadherin. Cadherins are divided into type I and type II subgroups. Type I cadherins include epithelial cadherin (E-cadherin, cadherin-1 or uvomorulin), neural cadherin (N-cadherin or cadherin-2), placental cadherin (P-cadherin or cadherin-3) and retinal cadherin (R-cadherin or cadherin-4). Kidney cadherin (K-cadherin or cadherin-6) and osteoblast cadherin (OB-cadherin or cadherin-11) are type II cadherins. The progression of carcinomas is associated with the loss of epithelial morphology and a concomitant acquisition of a more mesenchymal phenotype, which is thought to contribute to the invasive and/or metastatic behavior. A putative role for cadherin-11 in these late stages of tumor progression is based on the fact that migration of mesenchymal cells is facilitated when cadherin-11 is highly expressed.

Immunogen

Epitope: Cadherin 5 domain
KLH-conjugated linear peptide corresponding to the Cadherin 5 domain of Human Cadherin-11/OB Cadherin.

Application

Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Cadherin-11/OB Cadherin Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of Cadherin-11/OB Cadherin.
Western Blotting Analysis: 2 µg/mL of a representative lot of this antibody detected Cadherin-11/OB Cadherin in 10 µg of Mouse Brain, Mouse Brain Cytosol, Mouse Brain Membrane, Mouse Brain Nuclear Extract, Mouse (Aged) Brain Frontal Cortex, and Mouse Cerebellum tissue lysate.

Immunohistochemistry Analysis: A 1:50 dilution of a representative lot of this antibody detected Cadherin-11/OB Cadherin in neurons of the human cerebral cortex and thalamus, as well as purkinje and basket cells in human cerebellum.

Quality

Evaluated by Western Blotting in Human Thalamus Tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Cadherin-11/OB Cadherin in 10 µg of Human Thalamus Tissue lysate.

Target description

~75 kDa observed.
Full length protein has a molecular weight of ~120 kDa, however the cleaved product has a molecular weight of ~75-80 kDa (McCusker, C., et al. (2009) MBC. 20(1):78-89). Uncharacterized bands may be observed in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service