Skip to Content
Merck
All Photos(1)

Key Documents

MABT20

Sigma-Aldrich

Anti-OL-protocadherin Antibody, clone 5G10

clone 5G10, from rat

Synonym(s):

protocadherin 10, ortholog of OL-pcdh

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$743.00

$743.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$743.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$743.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5G10, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Pcdh10(18526)

General description

OL-protocadherin (OL-pc, protocadherin 10) is a transmembrane protein and a member of the cadherin superfamily, which is characterized by its members’ abilities to undergo interactions that are homophilic in nature via their extracellular domain. Several protocadherins have been observed within the nervous system, and a few have been shown to localize at synapses. With OL-pc, the homophilic binding has been shown to stimulate cell aggregation when the protein is introduced into cadherin-deficient cells. Although the function of OL-protocadherin has yet to be elucidated, studies have shown OL-pc to be necessary to axonal normal projection through the ventral telencephalon, where OL-pc shows highest expression at the striatal region.

Specificity

The antibody recognizes the cytoplasmic domain of OL-protocadherin.

Immunogen

Epitope: Cytoplasmic domain
GST-tagged recombinant protein corresponding to the cytoplasmic domain of mouse OL-protocadherin.

Application

Anti-OL-protocadherin Antibody, clone 5G10 is an antibody against OL-protocadherin for use in WB, IC, IH.
Immunofluorescence Analysis: The antibody has been shown to detect OL-protocadherin in IF. (Nakao, 2008).

Immunohistochemistry Analysis: The antibody has been shown to detect OL-protocadherin in IH. (Aoki, 2003).
Research Category
Cell Structure

Neuroscience
Research Sub Category
Adhesion (CAMs)

Synapse & Synaptic Biology

Quality

Evaluated by Western Blot in mouse E13-E14 brain tissue lysate.

Western Blot Analysis: 1 µg/mL of the antibody detected OL-protocadherin in 10 µg of mouse E13-E14 brain tissue lysate.

Target description

~ 130 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine, pH 7.4, 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse E13-E14 brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khoa Tran-Anh et al.
eNeuro, 7(6) (2020-10-16)
One of the notable characteristics of the functional localization in the cerebellar cortex is the dual representation of the body (somatotopy) on its anterior-posterior axis. This somatotopy is conspicuous in the C1/C3 module, which is demarcated as the multiple zebrin-negative
Joyce M Hoek et al.
PeerJ, 8, e9300-e9300 (2020-06-18)
Despite the widespread use of antibodies as a research tool, problems with specificity, lot-to-lot consistency and sensitivity commonly occur and may be important contributing factors to the 'replication crisis' in biomedical research. This makes the validation of antibodies and accurate
Naoki Ihara et al.
The European journal of neuroscience, 44(3), 1998-2003 (2016-05-22)
In the mouse olfactory system, the axons of olfactory sensory neurons that express the same type of odorant receptor (OR) converge to a specific set of glomeruli in the olfactory bulb (OB). It is widely accepted that expressed OR molecules
Suteera Vibulyaseck et al.
The Journal of comparative neurology, 525(14), 2971-2990 (2017-05-26)
Transversely oriented lobules and longitudinally arrayed stripes of Purkinje cell subsets subdivide the cerebellar cortex into multiple compartments that are involved in diverse functions. In the mammalian cerebellum, anterior, and posterior lobules, which are involved in somatosensorimotor function, show an
Yuanjun Luo et al.
Scientific reports, 13(1), 7114-7114 (2023-05-03)
The olivocerebellar projection is organized into an intricate topographical connection from the inferior olive (IO) subdivisions to the longitudinally-striped compartments of cerebellar Purkinje Cells (PCs), to play an essential role in cerebellar coordination and learning. However, the central mechanisms for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service