Merck
  • Spatial rearrangement of Purkinje cell subsets forms the transverse and longitudinal compartmentalization in the mouse embryonic cerebellum.
Spatial rearrangement of Purkinje cell subsets forms the transverse and longitudinal compartmentalization in the mouse embryonic cerebellum.
The Journal of comparative neurology (2017-05-26)
Suteera Vibulyaseck, Hirofumi Fujita, Yuanjun Luo, Anh Khoa Tran, Arata Oh-Nishi, Yuichi Ono, Shinji Hirano, Izumi Sugihara
ABSTRACT
Transversely oriented lobules and longitudinally arrayed stripes of Purkinje cell subsets subdivide the cerebellar cortex into multiple compartments that are involved in diverse functions. In the mammalian cerebellum, anterior, and posterior lobules, which are involved in somatosensorimotor function, show an alternation of aldolase C (zebrin II) -positive and -negative stripes, whereas the central lobules (lobules VIb-VII and crus I), which are implicated in nonmotor functions, show a laterally expanded arrangement solely of aldolase C-positive stripes. To understand the developmental process of this compartmental pattern, we identified groups of Purkinje cell subsets in the entire mouse cerebellum at embryonic day (E) 14.5 by staining Purkinje cell subset markers. We then tracked four major domains of Protocadherin 10 (Pcdh10)-positive Purkinje cell subsets (medial, dorsal, central, and mid-lateral subsets), which were clearly demarcated during E14.5-17.5. These domains of Purkinje cell subsets shifted predominantly in the longitudinal direction to be positioned in the anterior and posterior lobules. However, a particular portion of the medial and mid-lateral domains, and the whole of the central domain shift in the lateral direction to be positioned in the central lobules. The results indicate that while the longitudinal shift of domains of Purkinje cell subsets forms the longitudinally striped compartments in the anterior and posterior cerebellum, the lateral shift of particular domains of Purkinje cell subsets underlies the laterally expanded arrangement of stripes in central lobules. Thus, the rearrangement of Purkinje cell subsets in the embryonic cerebellum is critically related to the compartmental organization in the mammalian cerebellum.
Sigma-Aldrich
Anti-OL-protocadherin Antibody, clone 5G10, clone 5G10, from rat
Pricing and availability is not currently available.