Skip to Content
Merck
All Photos(1)

Key Documents

MABS2041

Sigma-Aldrich

Anti-pan-ARF Antibody, clone 1D9

clone 1D9, from mouse

Synonym(s):

ADP-ribosylation factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$252.00
100 μG
$647.00

$252.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$252.00
100 μG
$647.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$252.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D9, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
electron microscopy: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ARF1(375)

General description

ADP-ribosylation factor 1 (UniProt: P84077; also known as Arf1) is encoded by the ARF1 gene (Gene ID: 375) in human. Arf1 is a member of the family of low molecular weight GTP-binding proteins. It is abundant in neural tissues where it may comprise up to 1% of total cellular protein. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. Arf1, 3, 4, and 5 are predominantly cytosolic, but could be recruited to a variety of intracellular membranes, but not plasma membranes, upon incubation in the presence of GTP S. Arf6 is found at the plasma membrane and in endosomes. Arf1 is localized to the Golgi apparatus and has a central role in intra-Golgi transport. It has two nucleotide binding regions (aa 24-32 and 126-129). Arf1 functions as an allosteric activator of the cholera toxin catalytic subunit, an ADP-ribosyltransferase. It is also involved in protein trafficking among different compartments and is reported to modulate vesicle budding and uncoating within the Golgi complex. In its GTP-bound form, its triggers the association with coat proteins with the Golgi membrane. The hydrolysis of Arf1-bound GTP, which is mediated by ARFGAPs proteins, is required for dissociation of coat proteins from Golgi membranes and vesicles. This antibody (clone 1D9) detects all Arf proteins, but to different degrees. (Ref.: Cavenagh, MM., et al. (1996). J. Biol. Chem. 271(36):21767-74).

Specificity

Clone 1D9 detects all isoforms of DP-ribosylation factors in human cells.

Immunogen

Purified full length human recombinant ADP-ribosylation factor 1.

Application

Anti-pan-ARF, clone 1D9, Cat. No. MABS2041, is a mouse monoclonal antibody that detects all ADP-ribosylation factors and has been tested for use in ELISA, Electron Microscopy, Immunoprecipitation, and Western Blotting,
Research Category
Signaling
Western Blotting Analysis: 1 ug/mL from a representative lot detected ARF proteins in human lung tissue lysate.

Immunoprecipitation Analysis: A representative lot immunoprecipitated ARF proteins (Cavenagh, M.M., et. al. (1996). J Biol Chem. 271(36):21767-74).

ELISA Analysis: A representative lot detected pan-ARF in ELISA applications (Cavenagh, M.M., et. al. (1996). J Biol Chem. 271(36):21767-74).

Western Blotting Analysis: A representative lot detected ARF proteins in Western Blotting applications (Cavenagh, M.M., et. al. (1996). J Biol Chem. 271(36):21767-74; Zuezem, S., et. al. (1992). Proc Natl Acad Sci USA. 89(14):6619-23).

Electron Microscopy Analysis: A representative lot detected ARF proteins in Electron Microscopy applications (Zuezem, S., et. al. (1992). Proc Natl Acad Sci USA. 89(14):6619-23).

Dot Blot Analysis: A representative lot detected ARF proteins in Dot Blot applications (Cavenagh, M.M., et. al. (1996). J Biol Chem. 271(36):21767-74).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 ug/mL of this antibody detected pan-ARF in HeLa cell lysate.

Target description

~20 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service