Skip to Content
Merck
All Photos(2)

Key Documents

MABS2024

Sigma-Aldrich

Anti-Acetyl CoA Carboxylase 1 Antibody, clone 10H2.1

clone 10H2.1, from mouse

Synonym(s):

ACC1, ACC-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$214.00
100 μG
$545.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$214.00
100 μG
$545.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

10H2.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ACACA(31)

General description

Acetyl-CoA carboxylase 1 (UniProt: Q13085; also known as ACC1, ACC-alpha) is encoded by the ACACA (also known as ACAC, ACC1, ACCA) gene (Gene ID: 31) in human. It catalyzes the rate-limiting reaction in the biogenesis of long-chain fatty acids. ACC1 is a multi-subunit, biotin-dependent enzyme that catalyzes carboxylation of acetyl-CoA to produce malonyl-CoA through its two catalytic activities, biotin carboxylase (BC) and carboxyltransferase (CT). The activity of ACC can be controlled at the transcriptional level as well as by small molecule modulators and covalent modification. Two isoforms fof ACC have been reported, alpha and beta that are encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. ACC1 levels are regulated under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. ACC1 activity is inhibited upon phosphorylation, which occurs when glucagon or epinephrine binds to the receptors or when the energy status of the cell is low, leading to the activation of the AMP-activated protein kinase. The presence of fatty acids inhibits the activities of ACC1. ACC is considered as an important target in the design of new anti-obesity drugs.

Specificity

Clone 10H2.1 detects human Acetyl-CoA carboxylase 1. It targets an epitope within 245 amino acids from the C-terminal end.

Immunogen

GST-tagged recombinant fragment corresponding to 245 amino acids from the C-terminal region of human Acetyl-CoA carboxylase 1.

Application

Anti-Acetyl CoA Carboxylase 1, clone 10H2.1, Cat. No. MABS2024, is a mouse monoclonal antibody that detects Acetyl-CoA carboxylase 1 and has been tested for use in Western Blotting and Immunohistochemistry (Paraffin).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Acetyl CoA Carboxylase 1 in A431 cell lysate.

Target description

~265 kDa observed; 265.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service