Skip to Content
Merck
All Photos(1)

Key Documents

MABS14

Sigma-Aldrich

Anti-mTOR Antibody, clone F11

clone F11, from mouse

Synonym(s):

FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP-rapamycin associated protein, FKBP12-rapamycin complex-associated prot

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$683.00

$683.00


Estimated to ship on09 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$683.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$683.00


Estimated to ship on09 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F11, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MTOR(2475)

General description

mTOR (Mammalian Target of Rapamycin, aka FRAP, RAPT or RAFT) is a large 289 kDa Ser/Thr protein kinase that regulates cell cycle progression, cell growth, protein synthesis, ribosome biogenesis, and autophagy. mTOR is an evolutionarily conserved member of the Phosphoinositol Kinase-related Kinase (PIKK) family whose activity is regulated by phosphorylation on Ser2448 by Akt in response to insulin or muscle activity. mTORC1 (mTOR Complex 1) consists of at least mTOR, Raptor, and G (mLST8). mTORC1 is known to play a central role in insulin signaling, which is crucial in maintaining metabolic homeostasis. mTORC2 affects cell proliferation and survival primarily by phosphorylating the hydrophobic motif of Akt on Ser473, a well-known effecter of the PI3 Kinase pathway. In addition to phosphorylating Akt, the mTORC2 complex is also known to effect cytoskeletal organization and migration by exerting its effects through Rac, Rho, and PKC. Defects in both mTOR complexes are associated with a variety of diseases, including cancer and diabetes.

Immunogen

GST-tagged recombinant protein corresponding to rat mTOR.

Application

Immunoprecipitation Analysis: A previous lot was used by an independent laboratory in IP. (Nemazanyy, I, et al. (2008). Hybridoma, 27:395-398.)
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling
This Anti-mTOR Antibody, clone F11 is validated for use in WB, IP for the detection of mTOR.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected mTOR on 10 µg of HeLa cell lysate.

Target description

~ 289 kDa observed

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Habeebunnisa Begum et al.
Drug development research, 83(4), 910-926 (2022-01-30)
Pancreatic cancer (PC) is among the most lethal cancers and is resistant to existing therapies, which highlights the need for new and alternative therapeutic treatments. Autophagy is emerging as one of the alternative cell death mechanisms and is well known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service