Skip to Content
Merck
All Photos(1)

Key Documents

09-345

Sigma-Aldrich

Anti-phospho-mTOR (Thr2446) Antibody

Upstate®, from rabbit

Synonym(s):

FRAP1, FRAP, FRAP2, RAFT1, RAPT1, FK506 binding protein 12-rapamycin associated protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$660.00

$660.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$660.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$660.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr2446)

Gene Information

human ... MTOR(2475)

General description

mTOR (Mammalian Target of Rapamycin, aka FRAP, RAPT or RAFT) is a large 289 kDa Ser/Thr protein kinase that regulates cell cycle progression, cell growth, protein synthesis, ribosome biogenesis, and autophagy. mTOR is an evolutionarily conserved member of the Phosphoinositol Kinase-related Kinase (PIKK) family whose activity is regulated by phosphorylation on Ser2448 by Akt in response to insulin or muscle activity. Interestingly, mTOR is the central component of two multimeric kinase complexes consisting of mTOR and numerous other mTOR binding proteins. These two multimeric protein complexes are designated mTORC1 and mTORC2. mTORC1 (mTOR Complex 1) consists of at least mTOR, Raptor, and GβL (mLST8). mTORC1 is known to play a central role in insulin signaling, which is crucial in maintaining metabolic homeostasis. The complex is activated primarily though the PI3 Kinase/Akt pathway. Upon insulin stimulation, Akt activates mTORC1 by phosphorylating and inhibiting TSC (and possibly other yet discovered targets and/or mTOR itself). This inhibits the upstream small GTPase regulator Rheb (Ras homolog enriched in brain). This inhibits the kinase activity of the mTORC1 complex, thus disabling its ability to phosphorylate its downstream targets such as p70 S6K on Thr389 and 4E-BP1 on Thr229. The other mTOR complex, mTORC2 (mTOR Complex 2), is made up of at least mTOR, Rictor, GL, Sin1, Protor 1 and 2. mTORC2 affects cell proliferation and survival primarily by phosphorylating the hydrophobic motif of Akt on Ser473, a well-known effecter of the PI3 Kinase pathway. In addition to phosphorylating Akt, the mTORC2 complex is also known to effect cytoskeletal organization and migration by exerting its effects through Rac, Rho, and PKC. Interestingly, unlike mTORC1, the mTORC2 complex appears to not be inhibited by treatment with rapamycin and for this reason is referred to as the rapamycin-insensitive complex. Defects in both mTOR complexes are associated with a variety of diseases, including cancer and diabetes.

Specificity

Recognizes mTOR when phosphorylated on Thr2446

Immunogen

Amino acids encompassing and including phosphorylated Thr2446 of human mTOR

Application

Anti-mTOR Antibody detects level of phospho-mTOR (Thr2446) & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Glucose/Glycogen Metabolism

PI3K, Akt, & mTOR Signaling

Quality

Routinely evaluated by western blot on NGF-treated PC12 cell lysate.

Target description

289 kDa

Physical form

Affinity purified rabbit IgG in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH7.4.

Storage and Stability

Stable for 1 year at 2 to 8°C from date of shipment.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojie Gan et al.
Frontiers in immunology, 10, 2945-2945 (2020-01-31)
Hepatic ischemia/reperfusion injury (HIRI) is a major cause of liver dysfunction and even liver failure after liver transplantation and hepatectomy. One of the critical mechanisms that lead to HIRI is an acidic microenvironment, which develops due to the accumulation of
Johannes J Kovarik et al.
iScience, 26(1), 105717-105717 (2022-12-13)
To investigate long COVID-19 syndrome (LCS) pathophysiology, we performed an exploratory study with blood plasma derived from three groups: 1) healthy vaccinated individuals without SARS-CoV-2 exposure; 2) asymptomatic recovered patients at least three months after SARS-CoV-2 infection and; 3) symptomatic
Youyou Shao et al.
Journal of cellular physiology, 234(4), 4068-4080 (2018-08-28)
Asthma is characterized by chronic inflammation, and long-term chronic inflammation leads to airway remodeling. But the potential regulatory mechanism of airway remodeling is not clearly understood, and there is still no effective way to prevent airway remodeling. Present studies have
R Michelle Saré et al.
Frontiers in neuroscience, 16, 834890-834890 (2022-07-15)
Sleep is critical for brain development and synaptic plasticity. In male wild-type mice, chronic sleep restriction during development results in long-lasting impairments in behavior including hypoactivity, decreased sociability, and increased repetitive behavior. Disordered sleep is characteristic of many neurodevelopmental disorders.
LiJun Xu et al.
Molecular oncology, 12(3), 269-286 (2017-09-03)
To investigate whether IL-6 signaling affects the susceptibility of castration-resistant prostate cancer (CRPC) cells to cytotoxic action of natural killer (NK) cells, CRPC cell lines (having different IL-6 levels) were developed by lentiviral transduction. While observing no secreted IL-6 level

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service