Skip to Content
Merck
All Photos(3)

Key Documents

MABS1166

Sigma-Aldrich

Anti-AdipoR2 Antibody, clone 4D3.2

clone 4D3.2, from mouse

Synonym(s):

Adiponectin receptor protein 2, AdipoR2, Progestin and adipoQ receptor family member II

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$823.00

$823.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$823.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$823.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4D3.2, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ADIPOR2(79602)

General description

Adiponectin receptor protein 2 (UniProt Q86V24; also known as Progestin and adipoQ receptor family member II) is encoded by the ADIPOR2 (also known as PAQR2) gene (Gene ID 79602) in human. Adiponectin receptor protein 1 & 2 (AdipoR1 & AdipoR2) are receptors for adiponectin, a well knonwn anti-inflammatory adipokine highly abundant in serum. Both AdipoR1 and AdipoR2 are reported to be ubiquitously expressed with the highest expression in skeletal muscle and liver, respectively. Circulating adiponectin levels are reduced in obesity and type II diabetes, whereas weight loss elevates serum adiponectin. Adiponectin deficiency in mice is reported to exacerbate metabolic dysfunction by diet-induced obesity in some studies, while adiponectin overexpression alleviates metabolic dysfunction independent of weight loss in both genetic and diet-induced animal models of obesity. Adipor1-knockout (KO) results in increased adiposity and decreased glucose clearance in mice, whereas Adipor2-KO mice are lean and displayed improved glucose clearance. In addition to its metabolic role, adiponectin deficiency is associated with various clinical cardiovascular disorders including myocardial infarction, peripheral artery disease, and endothelial dysfunction. In a study of murine hind limb ischemia model, blood flow recovery after chronic hind limb ischemic surgery is similar among wild-type and AdipoR1-KO mice, while revascularization in AdipoR2-KO mice is severely attenuated. Human AdipoR2 is a 7-transmembrane (a.a. 148-168, 182-202, 214-234, 246-266, 274-294, 310-330, 349-369) protein with the opposite topology of G protein-coupled receptors (GPCRs), having its N-terminal end at the cytoplasmic side (a.a. 1-147) and C-terminal end at the extracellular side (a.a. 370-386). Unlike GPCRs also, adipoR1 & AdipoR2 transmembrane helices are not kinked or tilted relative to the plane of the membrane.

Specificity

Clone 4D3.2 targets an epitope within the N-terminal cytoplasmic domain.

Immunogen

Epitope: N-terminal cytoplasmic domain.
GST-tagged recombinant human AdipoR2 N-terminal fragment.

Application

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected AdipoR2 in human prostate cancer, as well as normal rat heart and human cerebral cortex tissue sections.
Research Category
Signaling
Research Sub Category
Lipid Metabolism & Weight Regulation
This Anti-AdipoR2 Antibody, clone 4D3.2 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of AdipoR2.

Quality

Evaluated by Western Blotting of GST-tagged human AdipoR2 N-terminal fragment.

Western Blotting Analysis: A 1:1000 dilution of this antibody detected 50 ng of GST-tagged recombinant human AdipoR2 N-terminal fragment.

Target description

43.88 kDa calculated.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangxi Sun et al.
Neoplasia (New York, N.Y.), 21(9), 921-931 (2019-08-12)
The survival of patients diagnosed with metastatic renal cell carcinoma (RCC) is still limited and the current targeted therapies are only partially effective. Herein, we investigated the clinical value and functions of adiponectin receptors (AdipoR1 and AdipoR2) in metastatic renal
Kiyoharu J Miyagishima et al.
Communications biology, 4(1), 1360-1360 (2021-12-11)
Late-onset retinal degeneration (L-ORD) is an autosomal dominant disorder caused by a missense substitution in CTRP5. Distinctive clinical features include sub-retinal pigment epithelium (RPE) deposits, choroidal neovascularization, and RPE atrophy. In induced pluripotent stem cells-derived RPE from L-ORD patients (L-ORD-iRPE)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service