Skip to Content
Merck
All Photos(1)

Key Documents

MAB8262X

Sigma-Aldrich

Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H, Alexa Fluor 488

clone 133/1H, Chemicon®, from mouse

Synonym(s):

RSV Fusion

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

133/1H, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2aκ

shipped in

wet ice

Specificity

Directed against the 47-49 kDa Fusion Protein of RSV. Antibody MAB858-1 recognizes both A and B RSV strains.

Immunogen

Respiratory Syncytial virus A2

Application

Detect Respiratory Syncytial Virus using this Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H, Alexa Fluor 488 validated for use in IF.
Immunofluorescence

Optimal working dilutions must be determined by the end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Purified, Alexa 488 conjugated. Liquid 0.02MPB, 0.25M NaCl, 0.1% sodium azide

Storage and Stability

Store at 2 to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rémi Villenave et al.
Virology journal, 8, 43-43 (2011-01-29)
Human respiratory syncytial virus (RSV) causes severe respiratory disease in infants. Airway epithelial cells are the principle targets of RSV infection. However, the mechanisms by which it causes disease are poorly understood. Most RSV pathogenesis data are derived using laboratory-adapted
Mechanism of action for respiratory syncytial virus inhibitor RSV604.
Challa, S; Scott, AD; Yuzhakov, O; Zhou, Y; Tiong-Yip, CL; Gao, N; Thresher, J; Yu, Q
Antimicrobial agents and chemotherapy null
Olivier Touzelet et al.
Molecular & cellular proteomics : MCP, 19(5), 793-807 (2020-02-23)
The respiratory epithelium comprises polarized cells at the interface between the environment and airway tissues. Polarized apical and basolateral protein secretions are a feature of airway epithelium homeostasis. Human respiratory syncytial virus (hRSV) is a major human pathogen that primarily
Ricardo de Souza Cardoso et al.
Viruses, 13(2) (2021-02-06)
Human respiratory syncytial virus (HRSV) is the most frequent cause of severe respiratory disease in children. The main targets of HRSV infection are epithelial cells of the respiratory tract, and the great majority of the studies regarding HRSV infection are
Ricardo S Cardoso et al.
mBio, 11(5) (2020-10-01)
Human respiratory syncytial virus (HRSV) envelope glycoproteins traffic to assembly sites through the secretory pathway, while nonglycosylated proteins M and N are present in HRSV inclusion bodies but must reach the plasma membrane, where HRSV assembly happens. Little is known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service