Skip to Content
Merck
All Photos(2)

Key Documents

MABS1164

Sigma-Aldrich

Anti-Nucleobindin-2/Nestafin-1 Antibody, clone 3B3.1

clone 3B3.1, from mouse

Synonym(s):

Nucleobindin-2, Nestafin-1, DNA-binding protein NEFA, Prepronesfatin, Pronesfatin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$803.00

$803.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$803.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$803.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B3.1, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Nucleobindin-2 (UniProt Q9JI85; also known as DNA-binding protein NEFA, Prepronesfatin) is encoded by the Nucb2 (also known as Nefa) gene (Gene ID 59295) in rat species. Nucleobindin-2 is initially translated as a 420-a.a. precursor protein. Cleavage of signal peptide (aa1-24) yields the 396-a.a. mature nucleobindin-2 with very high sequence homology among human, mouse, and rat species (more than 85%). Post-translational processing by prohormone convertases (PC 1/3 and PC 2) cleaves nucleobindin-2 into nesfatin-1, nesfatin-2, and nesfatin-3 (aa1–82, aa85–163, and aa166–396, respectively, numbering based on mature sequence). Nesfatin-1 is a well recognized circulating anorexigenic peptide, whose production increases post-prandially. Nucleobindin-2 contains two calcium-binding EF-hand domains separated by an acidic amino acid-rich region, and a leucine zipper. Nucleobindin-2 is known to be expressed in key hypothalamic nuclei and plays a role in energy homeostasis. Nucleobindin-2 is also expressed in various human peripheral tissues, including the stomach, pancreas, reproductive organs, and adipose tissues, suggesting roles in various metabolic functions. Nucleobindin-2 expression level is reported to be elevated in clear cell renal cell carcinoma (ccRCC), breast cancer, and prostate cancer (PCa) tissues.

Specificity

Clone 3B3.1 reacts with pro-nucleobindin-2, mature nucleobindin-2, and nesfatin-1, but not nesfatin-2 or nesfatin-3.

Immunogen

Epitope: Nesfatin-1
GST-tagged recombinant rat Nestafin-1.

Application

Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected nucleobindin-2/nestafin-1 immunoreactivity in human colon cancer tissue, as well as in normal testis, tonsil, and pancreas tissue sections.
Research Category
Signaling
Research Sub Category
Metabolic Hormones & Receptors
This Anti-Nucleobindin-2 Antibody, clone 3B3.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of Nestafin-1.

Quality

Evaluated by Western Blotting in mouse liver tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected nucleobindin-2 in 10 µg of mouse liver tissue lysate.

Target description

~50 kDa observed. 47.33/50.20 kDa (human mature/pro-nucleobindin-2), 47.42/50.30 kDa (mouse mature/pro-nucleobindin-2), 47.25/50.09 kDa (rat mature/pro-nucleobindin-2), 9.552/9.612/9.583 kDa (human/mouse/rat nesfatin-1) calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aristea Psilopanagioti et al.
Journal of neuroendocrinology, 32(9), e12899-e12899 (2020-09-10)
Feeding is a complex behaviour entailing elaborate interactions between forebrain, hypothalamic and brainstem neuronal circuits via multiple orexigenic and anorexigenic neuropeptides. Nucleobindin-2 (NUCB2)/nesfatin-1 is a negative regulator of food intake and body weight with a widespread distribution in rodent brainstem
Aristea Psilopanagioti et al.
International journal of molecular sciences, 23(23) (2022-12-12)
Data on animals emphasize the importance of the neuronal glucagon-like peptide-1 (GLP-1) receptor (GLP-1R) for feeding suppression, although it is unclear whether astrocytes participate in the transduction of anorectic GLP-1R-dependent signals. In humans, the brain circuitry underlying these effects remains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service