Skip to Content
Merck
All Photos(2)

Key Documents

MABC98

Sigma-Aldrich

Anti-Sphingosine 1-phosphate receptor 5 (S1P5) Antibody, clone 13F9.1

clone 13F9.1, from mouse

Synonym(s):

Sphingosine 1-phosphate receptor 5, S1P receptor 5, S1P5, Endothelial differentiation G-protein-coupled receptor 8, Sphingosine 1-phosphate receptor Edg-8, S1P receptor Edg-8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$801.00

$801.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$801.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$801.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13F9.1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (immunogen homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... S1PR5(53637)

Related Categories

General description

Sphingosine 1-phosphate receptor 5 (S1PR5 or S1P5), otherwise known as EDG8, is a G-protein coupled receptor which is activated by the lysosphingolipid sphingosine 1-phosphate (S1P). It is coupled to Gi/0 alpha and G12 signaling pathways. S1P5 is highly abundant in the brain, particularly in the corpus callosum, but S1P5 has also been detected in the spleen, placenta, lung, aorta, and peripheral blood leukocytes. In the brain, it regulates the transformation of radial glial cells into astrocytes, and the migration of oligodendrocyte precursor cells during development.

Immunogen

Recombinant protein corresponding to mouse Sphingosine 1-phosphate receptor 5 (S1P5).

Application

Detect Sphingosine 1-phosphate receptor 5 (S1P5) using this Anti-Sphingosine 1-phosphate receptor 5 (S1P5) Antibody, clone 13F9.1 validated for use in Western Blotting & IHC.
Immunohistochemistry Analysis: A 1:300 dilution from a representative lot detected Sphingosine 1-phosphate receptor 5 in normal human cerebellum and normal placenta tissue.

Quality

Evaluated by Western Blot in Daudi cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Sphingosine 1-phosphate receptor 5 in 10 µg of Daudi cell lysate.

Target description

~42 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service