Skip to Content
Merck
All Photos(2)

Key Documents

MABN681

Sigma-Aldrich

Anti-TrkA Antibody, clone 6B2

ascites fluid, clone 6B2, from mouse

Synonym(s):

High affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, TRK1-transforming tyrosine kinase protein, Tropomyosin-related kinase A, Tyrosine kinase receptor, Tyrosine kinase receptor A, Trk-A, gp140trk, p140-TrkA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$750.00

$750.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$750.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$750.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6B2, monoclonal

species reactivity

human, rat

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NTRK1(4914)

General description

TrkA, apparent molecular weight 140 kDa, is a high affinity nerve growth factor (NGF). The Trk proto-oncogene family contains four members, TrkA, TrkB, TrkC, and TrkE, which are variably expressed throughout the central and peripheral nervous systems. TrkA binds to nerve growth factor (NGF) and autophosphorylates on tyrosine residues (Tyr490, Tyr674, Tyr675, Tyr751 and Tyr785) to activate multiple downstream effector proteins. Phosphorylation at Tyr490 is required for Shc association and subsequent activation of the Ras-MAP kinase-signaling cascade, which leads to activation of Elk-1-dependent gene transcription and neurite growth. Phosphorylations at Tyr674 and Tyr675 lie within the catalytic domain of TrkA tyrosine kinase and reflect Trk kinase activity. Additionally, phosphorylation at Tyr751 is required for PI3-kinase association and activation of the Akt signaling cascade.

Immunogen

Purified recombinant extracellular fragment of TrkA expressed in HEK293 cells.

Application

Anti-TrkA Antibody, clone 6B2 is a highly specific mouse monoclonal antibody, that targets TrkA & has been tested in western blotting, IHC & ICC.
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected TrkA in human brain and rat cerebellum tissues.

Immunocytochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected TrkA in PC-12 cells.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected TrkA in 10 µg of human brain tissue lysate.

Target description

~87 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 4 isoforms ranging between ~77 kDa and ~87 kDa

Physical form

Mouse monoclonal IgG2b ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Human brain tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew D Johnson et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 39(2), 1010428317692256-1010428317692256 (2017-02-22)
This study investigated the Trk receptor family as a therapeutic target in pancreatic ductal adenocarcinoma and assessed their prognostic significance. Global gene expression analysis was investigated in prospectively collected pancreatic ductal adenocarcinomas that had either undergone neoadjuvant chemoradiation or were
Angels Almenar-Queralt et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(9), 3320-3339 (2014-02-28)
Overexpression and/or abnormal cleavage of amyloid precursor protein (APP) are linked to Alzheimer's disease (AD) development and progression. However, the molecular mechanisms regulating cellular levels of APP or its processing, and the physiological and pathological consequences of altered processing are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service