Skip to Content
Merck
All Photos(2)

Key Documents

MABE1008

Sigma-Aldrich

Anti-TRRAP Antibody, clone 2TRR-1B3

ascites fluid, clone 2TRR-1B3, from mouse

Synonym(s):

Transformation/transcription domain-associated protein, 350/400 kDa PCAF-associated factor, PAF350/400, STAF40, TR-AP, Tra1 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$803.00

$803.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$803.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$803.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2TRR-1B3, monoclonal

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TRRAP(8295)

General description

Transformation/transcription domain-associated protein (UniProt Q9Y4A5; also known as 350/400 kDa PCAF-associated factor, PAF350/400, STAF40, TR-AP, Tra1 homolog) is encoded by the TRRAP (also known as PAF400) gene (Gene ID 8295) in human. Transformation/transcription domain-associated protein (TRRAP) is a cofactor component shared by several histone acetyltransferase (HAT) complexes, including SAGA (Spt-Ada-Gcn5-acetyltransferase), PCAF (P300/CBP-associated factor), and Tip60. TRRAP is an essential component of the histone epigenetic modifying mechinery that plays an important role in regulating cell-cycle progression. Trrap gene-knockdown or protein downregulation causes cell and mouse lethality, impaired spindle checkpoint and differentiation.

Specificity

The immunogen sequence is present in both spliced isoforms of human TRRAP reported by UniProt (Q9Y4A5).

Immunogen

Epitope: N-terminal to nuclear localization signal in the TP53-interacting region.
Ovalbumin-conjugated linear peptide corresponding to a sequence from the TP53-interacting region of human TRRAP.

Application

Anti-TRRAP Antibody, clone 2TRR-1B3 is an antibody against TRRAP for use in Western Blotting, Immunoprecipitation.
Immunoprecipitation Analysis: 10 µL from a representative lot immunoprecipitated TRRAP from 1 mL (4.5 mg/mL) of HeLa nuclear extract (Courtesy of Dr. Mustapha OULAD-ABDELGHANI, IGBMC, France).
Immunoprecipitation Analysis: A representative lot immunodepleted TRRAP using nonhomologous end-joining/NHEJ-competent protein extracts prepared from human AAH-1 lymphoblastoid cells (Robert, F., et al. (2006). Mol. Cell. Biol. 26(2):402-412).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins

Quality

Evalulated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TRRAP in 10 µg of HeLa nuclear extract.

Target description

~400 kDa observed. 437.6 kDa (isoform 1) and 434.4 kDa (isoform 2) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Mouse monoclonal IgG2aκ ascites without preservatives.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service