Skip to Content
Merck
All Photos(2)

Documents

MABN318

Sigma-Aldrich

Anti-Transmembrane Prostatic Acid Phosphatase Antibody/TMPAP, clone 3G10.1

clone 3G10.1, from mouse

Synonym(s):

Prostatic acid phosphatase, PAP, 5′-nucleotidase, 5′-NT, Ecto-5′-nucleotidase, Thiamine monophosphatase, Transmembrane Prostatic Acid Phosphatase, TMPase, ACPP, TMPAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G10.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACPP(55)

General description

ACPP, also known as Prostatic acid phosphatase, or PAP, 5′-nucleotidase (5′-NT), Ecto-5′-nuclotidase, or Thiamine monophosphatase (TMPase) , and encoded by the gene ACPP, is a non-specific tyrosine phosphatase that has both a secreted form and a cell membrane bound form. ACPP dephosphorylates a diverse number of substrates including alkyl, aryl, and acyl orthophosphate monoesters and phosphorylated proteins. ACPP also has lipid phosphatase activity and inactivates lysophosphatidic acid in seminal plasma. Additionally, the membrane bound form of ACPP has ecto-5′-nucleotidase activity in dorsal root ganglion (DRG) neurons and acts as a pain suppressor. Finally ACPP can also act as a tumor suppressor of prostate cancer. ACPP isoform 1is secreted and isoform 2 is membrane bound. Isoform 2 appears to shuttle between the cell membrane, lysosomes, and intracellular vesicles. ACPP is expressed in many tissues and is used as a marker in various stages of prostatic cancer diagnostics. Also the enzyme was intensely studied in HIV infection because it can be proteolytically cleaved in seminal fluid to produce several peptides (SEVI peptides) one of which, PAPf39, happens to enhance HIV infection by binding HIV virions and helping to attach them to target cells--interestingly such SEVI peptides are degraded in vitro by EGCG, a compound found in green tea extract.

Specificity

This antibody is expected to cross react with isoform 2 based upon immunogen design.

Immunogen

GST-tagged recombinant protein corresponding to human Transmembrane Prostatic Acid Phosphatase/TMPAP.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Transmembrane Prostatic Acid Phosphatase/TMPAP in human prostate tissue.
Research Category
Neuroscience
This Anti-Transmembrane Prostatic Acid Phosphatase Antibody/TMPAP, clone 3G10.1 is validated for use in western blotting & IHC (Paraffin) for the detection of Transmembrane Prostatic Acid Phosphatase/TMPAP.

Quality

Evaluated by Western Blotting in human prostate tumor tissue lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected this antibody detected Transmembrane Prostatic Acid Phosphatase/TMPAP in 200 µg of human prostate tumor tisssue lysate.

Target description

~48 kDa observed. Two isoforms are known to exist at 45 kDa (isoform 1) and 48 kDa (isoform 2). Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Sui et al.
British journal of cancer, 125(7), 1003-1015 (2021-07-12)
Metastasis is the major cause of treatment failure and cancer-related deaths in prostate cancer (PCa) patients. Our previous study demonstrated that a CD44+ subpopulation isolated from PCa cells or tumours possesses both stem cell properties and metastatic potential, serving as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service