Skip to Content
Merck
All Photos(1)

Key Documents

MABN1598

Sigma-Aldrich

Anti-RetGC-1/GUCY2D Antibody, clone 6D8.2

clone 6D8.2, from mouse

Synonym(s):

Retinal guanylyl cyclase 1, Guanylate cyclase 2D, retinal, RETGC-1, Rod outer segment membrane guanylate cyclase, ROS-GC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$680.00

$680.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$680.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$680.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6D8.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GUCY2D(3000)

General description

Retinal guanylyl cyclase 1 (EC 4.6.1.2; UniProt Q02846; also known as Guanylate cyclase 2D, retinal, RETGC-1, Rod outer segment membrane guanylate cyclase, ROS-GC) is encoded by the GUCY2D (also known as CORD6, GUC1A4, GUC2D, RETGC, RETGC1) gene (Gene ID 3000) in human. RetGC-1 is a membrane-bound retinal guanylyl cyclase expressed predominantly in the cone and rod photoreceptors in the cone outer segment. RetGC-1 and the related RetGC-2 mediate the synthesis of cyclic 3′, 5′-guanosine monophosphate (cGMP) from guanosine triphosphate in mammalian photoreceptor cells, where RetGC-1 and its associated activator proteins are responsible for the Ca2+-sensitive restoration of cGMP levels after light activation of the phototransduction cascade. RetGC-1 is produced with a signal peptide (a.a. 1-51) sequence, the removal of which yields the mature enzyme with an exracellular domain (a.a. 52-462), followed by a transmembrane segment (a.a. 463-487) and a cytoplasmic tail (a.a. 488-1103) that contains a protein kinase homology domain (a.a. 525-808), a dimerization domain, and the guanylate cyclase catalytic domain (a.a. 880-1010). Heterozygous mutations in the GUCY2D gene have been shown to cause autosomal dominant cone and cone-rod dystrophies (adCODs and adCORDs, respectively), while homozygous or compound heterozygous mutations cause autosomal recessively inherited Leber Congenital Amaurosis (LCA), the most severe form of inherited retinopathy, resulting in total blindness or greatly impaired vision at birth or in early infancy.

Specificity

Clone 6D8.2 targets an epitope in the cytoplasmic kinase domain of human retinal guanylyl cyclase 1 (RetGC-1) encoded by the GUCY2D gene.

Immunogen

Epitope: Kinase domain.
GST-tagged recombinant human RetGC-1/GUCY2D kinase domain fragmen.

Application

Research Category
Neuroscience
Research Sub Category
Sensory & PNS
This Anti-RetGC-1 Antibody, clone 6D8.2 is validated for use in Western Blotting for the detection of GUCY2D.

Quality

Evaluated by Western Blotting in Y79 human retinoblastoma cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected RetGC-1/GUCY2D in 10 µg of Y79 human retinoblastoma cell lysate.

Target description

~120 kDa observed. 114.7/120.1 kDa (mature/pro-form) calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service