Skip to Content
Merck
All Photos(1)

Key Documents

MABF3145

Sigma-Aldrich

Anti-Envelope glycoprotein B Antibody, HCMV Antibody, clone 27-156

Synonym(s):

gB

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$253.00
100 μG
$638.00

$253.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$253.00
100 μG
$638.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$253.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

27-156, monoclonal

mol wt

calculated mol wt 101.95 kDa
observed mol wt ~120-140 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable

isotype

IgG2bκ

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

vaccinia virus ... HA(3654615)

General description

Envelope glycoprotein B (UniProt: P13201; also known as gB) is encoded by the gB (also known as UL55) gene (Gene ID: 3077424) in human cytomegalovirus (HHV-5). Human cytomegalovirus (HCMV), a member of the β-Herpesviridae family, is a major cause of morbidity and mortality in organ transplant recipients and patients with AIDS. Its envelope glycoprotein B is reported to be essential for the infectivity and is considered to be one of the most immunogenic virus-encoded proteins. It facilitates the entry of HCMV entry into cells by binding receptors and mediating membrane fusion. Glycoprotein B is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off to produce the mature protein that contains a virion surface domain (aa 23-751), a transmembrane domain (aa 752-772), and an intra-virion domain (aa 773-907). The amino acid sequences 152-158 and 237-244 are critical for fusion and/or binding to host cell membrane. During virion morphogenesis, envelope glycoprotein B probably accumulates in the endosomes and trans-Golgi where secondary envelopment occurs. It is then transported to the cell surface from where it is endocytosed and is directed to the trans-Golgi network. The ectodomain of HCMV glycoprotein B forms rosette-like aggregates that are similar to rosettes formed by the EBV glycoprotein B ectodomain. Four exposed hydrophobic residues in its fusion loop are shown to be essential for rosette formation. A linear domain of glycoprotein B, designated at AD-1, represents a dominant antibody binding site on this protein and is also required for oligomerization of glycoprotein B. (Ref.: Sharma, S., et al. (2013). Virology. 435(2); 239-249; Britt, WJ., et al. (2005). J. Virol. 79(7); 4066-4079; Jean, F., et al. (2000). Proc. Natl. Acad. Sci. USA. 97(6); 2864-2869; Speckner, ., et al. (1999). J. Gen. Virol. 80(8); 2183-2191; Schoppel, K., et al. (1996). Virology. 216(1); 133-145).

Specificity

Clone 27-156 is a mouse monoclonal antibody that detects the envelope glycoprotein B in Human cytomegalovirus (HCMV). It targets an epitope within the antigenic domain 1 (AD1) within the C-terminal half.

Immunogen

Whole Human cytomegalovirus (HCMV).

Application

Quality Control Applications

Evaluated by Western Blotting with recombinant HCMV glycoprotein B.

Western Blotting Analysis (WB): A 1:200,000 dilution of this antibody detected recombinant HCMV glycoprotein B.

Tested applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated HCMV Envelope glycoprotein B in Immunoprecipitation applications (Reschke, M., et al. (1995). J Gen Virol. 76(Pt1); 113-22; Meyer, G.A., et al. (2000). J Biol Chem. 275(6); 3857-66; Vanarsdall, A.L., et al. (2008). J Virol. 82(23);11837-50).

Immunocytochemistry Analysis: A representative lot detected HCMV Envelope glycoprotein B in Immunocytochemistry applications (Reschke, M., et al. (1995). J Gen Virol. 76(Pt1); 113-22; Jean, F., et al. (2000). Proc Natl Acad Sci USA. 97(6); :2864-9; Meyer, G.A., et al. (2000). J Biol Chem. 275(6); 3857-66; Strive, T., et al. (2002). J Virol. 76(3); 1252-64).

Neutralizing: A representative lot neutralized Envelope glycoprotein B of HCMV. (Schoppel, K., et al. (1996). Virology. 216(1);133-45; Kropff, B., et al. (1997). J Gen Virol. 78(Pt8); 1999-2007; Vanarsdall, A.L., et al. (2008). J Virol. 82(23); 11837-50).

Immunofluorescence Analysis: A representative lot detected HCMV Envelope glycoprotein B in Immunofluorescence applications (Reschke, M., et al. (1995). J Gen Virol. 76(Pt1);113-22; Jean, F., et al. (2000). Proc Natl Acad Sci USA. 97(6); 2864-9; Meyer, G.A., et al. (2000). J Biol Chem. 275(6); 3857-66; Strive, T., et al. (2002). J Virol. 76(3); 1252-64; Vanarsdall, A.L., et al. (2008). J Virol. 82(23); 11837-50).

ELISA Analysis: A representative lot detected HCMV Envelope glycoprotein B in ELISA applications (Sharma, S., et al. (2013). Virology. 435(2); 239-49).

Western Blotting Analysis: A representative lot detected HCMV Envelope glycoprotein B in Western Blotting applications (Reschke, M., et al. (1995). J Gen Virol. 76(Pt1); 113-22; Schoppel, K., et al. (1996). Virology. 216(1); 133-45; Kropff, B., et al. (1997). J Gen Virol. 78(Pt8);1999-2007; Jean, F., et al. (2000). Proc Natl Acad Sci USA. 97(6); 2864-9; Sharma, S., et al. (2013). Virology. 435(2); 239-49; Khanh Le-Trilling, V. T., et al. (2020). Cell Rep. 30(7); 2248-2260).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Physical form

Purified mouse monoclonal antibody IgG2b in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service