Skip to Content
Merck
All Photos(1)

Key Documents

MABF220

Sigma-Aldrich

Anti-mA20 Antibody/TNFAIP3, clone 37

clone 37, 0.5 mg/mL, from rat

Synonym(s):

Tumor necrosis factor alpha-induced protein 3, TNF alpha-induced protein 3, Putative DNA-binding protein A20, Zinc finger protein A20

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$481.00

$481.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$481.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$481.00


Estimated to ship on14 April 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

37, monoclonal

species reactivity

mouse

concentration

0.5 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Tnfaip3(21929)

General description

TNFAIP3/mA20, also known as Tumor necrosis factor alpha-induced protein 3, OTU domain-containing protein 7C, Putative DNA-binding protein A20, or Zinc finger protein A20, and encoded by the gene TNFAIP3/OTUD7C, is an ubiquitin editing enzyme that contains both ubiquitin ligase and deubiquitinase activities. TNFAIP3/mA20 is involved in immune and inflammatory responses signaled by cytokines or pathogens through the Toll-like receptors and ultimately regulating NF-kappa-B activity. TNFAIP3/mA20 is an essential component of the ubiquitin editing protein complex consisting of RNF11, ICTH, and TAX1BP1 and ensures the transient nature of inflammatory signaling. TNFAIP3/mA20 is also a key player in switching of Lys-63 polyubiquitinated chains to Lys-48 polyubiquitinated chains for the subsequent degradation of specific targets as well. TNFAIP3/mA20 is located in the cytoplasm but can be found in the nucleus and lysosome depending upon the microenvironment and stimulus.

Immunogen

Recombinant protein corresponding to Mouse mA20/TNFAIP3.

Application

Detect mA20/TNFAIP3 using this Anti-mA20 Antibody/TNFAIP3, clone 37 validated for use in flow cytometry.
Research Category
Inflammation & Immunology
Research Sub Category
Apoptosis - Additional

Target description

90 kDa calculated

Physical form

Format: Purified
Purified rat monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service