Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200807

Sigma-Aldrich

Anti-Superoxide Dismutase (SOD) antibody, Mouse monoclonal

clone SD-G6, purified from hybridoma cell culture

Synonym(s):

Cu-Zn, Superoxide dismutase 1, hSod1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$286.00
100 μL
$759.00

$286.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$286.00
100 μL
$759.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$286.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

SD-G6, monoclonal

form

buffered aqueous solution

species reactivity

human, rat, canine

packaging

antibody small pack of 25 μL

technique(s)

indirect ELISA: 0.3-0.6 μg/mL using 5μg/ml Superoxide Dismutase from human erythrocytes for coating

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOD1(6647)

Related Categories

General description

Superoxide Dismutase (SOD) is a family of metalloenzymes widely distributed in both plants and animals. Superoxide dismutases appear to protect cells against reactive free radicals by scavenging the superoxide radicals produced by ionization radiation or through other mechanisms. SOD enzyme catalyzes the conversion of single electron reduced species of molecular oxygen to hydrogen peroxide and oxygen. There are several classes of SOD that differ in their metal binding ability, distribution in different cell compartments, and sensitivity to various reagents. Among these, Cu, Zn superoxide dismutase (SOD1) is widely distributed and comprises 90% of the total SOD. This ubiquitous enzyme, which requires Cu and Zn for its activity, has great physiological significance and therapeutic potential.

Immunogen

recombinant human copper-zinc superoxide dismutase (Cu-Zn-SOD).

Application

Monoclonal Anti-Superoxide Dismutase (SOD) recognizes natural (human erythrocyte SOD), recombinant SOD (human Cu-Zn-SOD and human placental SOD) and the enzymatically inactive form of these enzymes. Reactivity has been observed with SOD from human, rat and dog origin, no reactivity was observed with SOD from bovine, bacillus stearothermophilus or E. coli (Fe or Mn) origin. The antibody is recommended to use in various immunological techniques, including ELISA, Immunohistochemistry and immunofluorescence.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service