Skip to Content
Merck
All Photos(1)

Key Documents

MABF2182

Sigma-Aldrich

Anti-CD46 Antibody, clone TRA-2-10

clone TRA-2-10, from mouse

Synonym(s):

Membrane cofactor protein, TLX, Trophoblast leukocyte common antigen

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$252.00
100 μL
$666.00

$252.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$252.00
100 μL
$666.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$252.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

TRA-2-10, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CD46(4179)

General description

Membrane cofactor protein (UniProt: P15529; also known as TLX, Trophoblast leukocyte common antigen, CD46) is encoded by the CD46 (also known as MCP; MIC10) gene (Gene ID: 4179) in human. CD46 is a single-pass type I membrane protein that is expressed by all cells except erythrocytes. It acts as a cofactor for complement factor I, a serine protease that protects autologous cells against complement-mediated injury by cleaving C3b and C4b deposited on host tissue. It also acts as a costimulatory factor for T-cells which induces the differentiation of CD4+ into T-regulatory 1 cells. CD46 is synthesized with a signal peptide (aa 1-34), which is cleaved off in the mature form. The mature form contains an extracellular domain (aa 35-343), a transmembrane domain (aa 344-366), and a cytoplasmic domain (aa 367-392). It also contains four Sushi domains where Sushi domains 1 and 2 are required for interaction with human adenovirus B PIV/FIBER protein and with Measles Virus H protein, domains 2 and 3 are essential for Herpes virus 6 binding, and domains 3 and 4 are required for interaction with Streptococcus Pyrogens M protein and are the most important for interaction with C3b and C4b. CD46 can be glycosylated on Asn 83, 114, and 273 and N-glycosylation on Asn 114 and 273 are required for its cytoprotective function. N-glycosylation on Asn 114 is also required for Measles virus binding and N-glycosylation on Asn-273 is required for Neisseria binding. CD46 can also be extensively O-glycosylated in the Ser/Thr-rich domain, which is required for Neisseria binding, but not for Measles virus binding. Mutations in CD46 gene have been linked to hemolytic uremic syndrome atypical 2 that has been characterized by microangiopathic hemolytic anemia, thrombocytopenia, and renal failure.

Specificity

Clone TRA-2-10 specifically detects CD46 in human cells.

Immunogen

2102Ep human embryonal carcinoma cells.

Application

Anti-CD46, clone TRA-2-10, Cat. No. MABF2182, is a mouse monoclonal antibody that detects Membrane cofactor protein (CD46) and has been for use in Flow Cytometry, Immunoprecipitation, Immunohistochemistry, and Inhibition assay.
Inhibition Analysis: A representative lot blocked Measles virus infection in BC1 cells. (Manchester, M., et. al. (1995). Proc Natl Acad Sci U S A. 92(6):2303-7).

Immunohistochemistry Analysis: A representative lot detected CD46 in Immunohistochemistry applications (Kemper, C., et. al. (2001). Clin Exp Immunol. 124(2):180-9).

Immunoprecipitation Analysis: A representative lot immunoprecipitated CD46 in Immunoprecipitation applications (Andrews, P.W., et. al. (1985). Ann Hum Genet. 49(Pt 1):31-9).

Flow Cytometry Analysis: A representative lot detected CD46 in Flow Cytometry applications (Manchester, M., et. al. (1995). Proc Natl Acad Sci U S A. 92(6):2303-7).
Research Category
Inflammation & Immunology

Quality

Evaluated by Flow Cytometry in HeLa cells.

Flow Cytometry Analysis: 1 µg of this antibody detected CD46 in one million HeLa cells.

Target description

43.75 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Dear Team, what is the antibody concentration for the 25uL and the 100uL? Thank you

    1 answer
    1. The concentration is listed on the lot-specific Certificate of Analysis. Please see the link below to view a Certificate:
      https://www.emdmillipore.com/product/Anti-CD46-Antibody-clone-TRA-2-10,MM_NF-MABF2182-100UL#documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service