Skip to Content
Merck
All Photos(1)

Key Documents

MABF1033

Sigma-Aldrich

Anti-CCRL2 (Mouse) Antibody, clone BZ2E3

clone BZ2E3, from rat

Synonym(s):

C-C chemokine receptor-like 2, Chemokine receptor CCR11, G-protein coupled beta chemokine receptor, L-CCR, Lipopolysaccharide-inducible C-C chemokine receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$417.00

$417.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$417.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$417.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

BZ2E3, monoclonal

species reactivity

mouse

should not react with

human

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Ccrl2(54199)

Related Categories

General description

C-C chemokine receptor-like 2 (UniProt: O35457; also known as Chemokine receptor CCR11, G-protein coupled beta chemokine receptor, Lipopolysaccharide-inducible C-C chemokine receptor, L-CCR, CCRL2) is encoded by the Ccrl2 (also known as Ccr11, Lccr) gene (Gene ID: 54199) in murine species. CCRL2 is a seven transmembrane, G-protein coupled receptor that serves as a receptor for CCL19 and chemerin/RARRES2. It does not appear to be a signaling receptor, but may have a role in modulating chemokine-triggered immune responses by capturing and internalizing CCL19 or by presenting RARRES2 ligand to CMKLR1, a functional signaling receptor. CCRL2 is expressed by the majority of cell types of hematopoietic origin, including monocytes, macrophages, neutrophils, lymphocytes, and NK cells. In some cell types, its expression is upregulated by inflammatory stimuli. CCRL2 induction is shown to be dependent on NF- B and JAK/STAT signaling pathways. It is constitutively expressed by mast cells and is detected in bronchial epithelium in OVA-induced airway inflammation. It is reported to be up-regulated during dendritic cell (DC) maturation. It is also considered to be important for the development of Th2 responses and may act as a decoy receptor capturing and endocyting its ligands CCL5 and CCL19. (Ref.: Monnier, J. et al. (2012). J. Immunol. 189(2):956-967).

Specificity

Clone BZ2E3 reacts with mCCRL2, but not mCMKLR1, huCMKLR1, mCRTH2, huCCRL2, or mCCR10. Clone BZ2E3 immunostained the surface of mCCRL2-expressing cells, but not non-mCCRL2-expressing cells (Monnier, J., et al. (2012). J. Immunol. 189(2):956-967; Zabel, B.A., et al. (2008). J. Exp. Med. 205(10):2207-2220).

Immunogen

KLH-conjugated linear peptide corresponding to the N-terminal end sequence of mouse CCRL2 (Zabel, B.A., et al. (2008). J. Exp. Med. 205(10):2207-2220).

Application

Flow Cytometry Analysis: 10-100 µg/mL from a representative lot immunostained mouse peritoneal exudate cells from wild-type, but not Ccrl2-/- mice (Courtesy of Brian Zabel, Ph.D., Stanford University, U.S.A.).

Flow Cytometry Analysis: A representative lot immunostained liver and lung CD31+/CD146+ endothelial cells from wild-type, but not Ccrl2-/- mice. Intraperitoneal LPS injection prior to cells isolation upregulated surface CCRL2 staining on liver and lung CD31+/CD146+ endothelial cells (Monnier, J., et al. (2012). J. Immunol. 189(2):956-967).

Flow Cytometry Analysis: A representative lot detected cytokines-induced CCRL2 expression on the surface of bEND.3 mouse endothelioma cells. IKKbeta inhibitor BAY (11 7082) and/or JAK-1 inhibitor (sc-204021) inhibited cytokines-induced CCRL2 expression (Monnier, J., et al. (2012). J. Immunol. 189(2):956-967).

Flow Cytometry Analysis: A representative lot immunostained L1.2 mouse pre-B lymphoma cells expressing transfected mCCRL2, but not untransfected L1.2 or L1.2 transfectants expressing mCMKLR1, huCMKLR1, mCRTH2, huCCRL2, or mCCR10, nor non-CCRL2-expressing mouse blood cells. Chemerin, but not CCL2, blocked clone BZ2E3 staining of CCRL2-positive mouse peritoneal exudate cells (Zabel, B.A., et al. (2008). J. Exp. Med. 205(10):2207-2220).
Research Category
Inflammation & Immunology
This rat monoclonal Anti-CCRL2 (Mouse) Antibody, clone BZ2E3, Cat. No. MABF1033, is validated for use in Flow Cytometry for the detection of CCRL2.

Quality

Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be rat IgG2a .

Target description

40.75 kDa calculated.

Physical form

Format: Purified
Protein G purified.
Purified rat IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service