Skip to Content
Merck
All Photos(2)

Key Documents

MABE936

Sigma-Aldrich

Anti-ASF-1 Antibody, clone AK-96

clone AK-96, from mouse

Synonym(s):

Serine/arginine-rich splicing factor 1, Alternative-splicing factor 1, ASF-1, Splicing factor, arginine/serine-rich 1, pre-mRNA-splicing factor SF2, P33 subunit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$558.00

$558.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$558.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$558.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AK-96, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRSF1(6426)

General description

Alternative-splicing factor 1 (ASF-1), also known as SRp30a or SF2, is a member of the highly conserved family of SR proteins, a group of serine/arginine-rich domain-containing proteins that play a critical role in regulated and constitutive splicing of precursor mRNA. The SR family of proteins is characterized as having one to two RNA recognition motifs located in the n-terminal sequence, and an RS domain within the C-terminus that is rich in serine/arginine dipeptide repeats. ASF-1 has an inhibitory role in exon skipping, in order to maintain splicing accuracy and to monitor alternative splicing. Studies have shown that ASF-1 plays a role in the regulation of caspase-9 alternative splicing as well as being a critical component for ceramide to function as a mediator of alternative splicing for the same caspase.

Immunogen

Recombinant protein corresponding to human ASF-1.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-ASF-1 Antibody, clone AK-96 is validated for use in western blotting, IP & ICC for the detection of ASF-1.
Western Blotting Analysis: 1.0 µg/mL of this antibody detected ASF-1 in 10 µg of Raji and HepG2 cell lysate.
Immunocytochemistry Analysis: A representative lot detected ASF-1 in BJ fibroblast cells (Fregoso, O., et al. (2013). Mole. Cell. 50:56-66).
Immunoprecipitation Analysis: A representative lot immunoprecipitated ASF-1 in BJ fibroblast cell lysate (Fregoso, O., et al. (2013). Mole. Cell. 50:56-66).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected ASF-1 in 10 µg of HeLa cell lysate.

Target description

~30 kDa observed

Linkage

Replaces: MABE512

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Rivera et al.
Epigenetics & chromatin, 14(1), 51-51 (2021-11-26)
Nuclear processes such as transcription and RNA maturation can be impacted by subnuclear compartmentalization in condensates and nuclear bodies. Here, we characterize the nature of nuclear granules formed by REST corepressor 2 (RCOR2), a nuclear protein essential for pluripotency maintenance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service