Skip to Content
Merck
All Photos(1)

Key Documents

MABE893

Sigma-Aldrich

Anti-LSm1 Antibody, clone 5F3

clone 5F3, from rat

Synonym(s):

U6 snRNA-associated Sm-like protein LSm1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$493.00

$493.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$493.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$493.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F3, monoclonal

species reactivity

mouse

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Lsm1(67207)

General description

The protein named U6 snRNA-associated Sm-like protein LSm1 or Cancer associated Sm-like, or Small nuclear ribonuclear CaSm and encoded by the human gene LSM1/CASM is a critical binding protein that mediates replication dependent histone mRNA degradation during cell division. Lsm1 is part of the Lsm family of proteins. Lsm proteins are a ubiquitous family of proteins characterized by the Sm-domain. They exist as hexa- or heptameric RNA-binding complexes and carry out RNA-related functions. Lsm1 interacts directly with SLBP a histone stem-loop binding protein critical for histone mRNA processing and expression. Histone mRNAs do not contain introns or polyadenylation signals and are processed by direct endonucleolytic cleavage. Lsm1 binds directly to the 3’-terminal U-tract of U6 snRNA . Lsm1 is localized to the nucleus and is up regulated in many cancers including pancreatic cancer and many tumor cell lines.

Immunogen

Recombinant protein corresponding to mouse LSm1.

Application

Anti-LSm1 Antibody, clone 5F3 is an antibody against LSm1 for use in western blotting & IP.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Western Blotting Analysis: A representative lot from an independent laboratory detected LSm1 in MEF cell lysate (Hoefig, K. P., et al. (2012). Nat Struct Mol Biol. 20(1):73-81.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated LSm1 from MEF cell lysate (Hoefig, K. P., et al. (2012). Nat Struct Mol Biol. 20(1):73-81.).

Quality

Evaluated by Western Blotting in L-929 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected LSm1 in 10 µg of L-929 cell lysate.

Target description

~15 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poonam Dhillon et al.
Journal of virology, 92(24) (2018-09-28)
Rotavirus replicates in unique virus-induced cytoplasmic inclusion bodies called viroplasms (VMs), the composition and structure of which have yet to be understood. Based on the analysis of a few proteins, earlier studies reported that rotavirus infection inhibits stress granule (SG)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service