Skip to Content
Merck
All Photos(1)

Key Documents

MABE878

Sigma-Aldrich

Anti-RNA Helicase A/Dhx9 Antibody, clone 8E3

clone 8E3, from rat

Synonym(s):

ATP-dependent RNA helicase A, RHA, DEAH box protein 9, mHEL-5, Nuclear DNA helicase II, NDH II

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$537.00

$537.00


Estimated to ship on28 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$537.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$537.00


Estimated to ship on28 April 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

8E3, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DHX9(1660)

General description

RNA Helicase A, also known as DEAH box protein 9 (mHEL-5), DHX9, or Nuclear DNA helicase II (NDH II), and encoded by the mouse gene Dhx9, is a component of the CRD-mediated mRNA stability complex. The CRD-mediated complex is a protein complex that binds to, and promotes stabilization of, mRNA molecules containing the coding region instability determinant (CRD). In humans, it consists of IGF2BP1 and at least four additional proteins: HNRNPU, SYNCRIP, YBX1, and RNA Helicase A. RNA Helicase A operates by unwinding dsDNA and RNA in a 3′ to 5′ direction. Additionally, RNA Helicase A acts as a transcriptional activator and interacts with LARP6 which is a protein that has been associated with fasting glucose traits, type 2 diabetes and obesity as well as the stabilization of collagen mRNAs for CO1A1 and CO1A2. RNA Helicase A is localized to the nucleolus within the nucleus but can also be found in the cytoplasm localized in cytoplasmic mRNP granules containing untranslated mRNAs and thus the protein shuttles between the nucleus and cytoplasm depending upon the microenvironment.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of mouse RNA Helicase A/Dhx9.

Application

Research Category
Epigenetics & Nuclear Function
This Anti-RNA Helicase A/Dhx9 Antibody, clone 8E3 is validated for use in western blotting & ICC for the detection of RNA Helicase A/Dhx9.
Western Blotting Analysis: A representative lot detected RNA Helicase A/Dhx9 in NIH3T3 cell lysate (Kotani, M., et al. (2010). Hybridoma. 29(3):259-261).
Immunocytochemistry Analysis: A representative lot detected RNA Helicase A/Dhx9 in NIH3T3 cells (Kotani, M., et al. (2010). Hybridoma. 29(3):259-261).

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected RNA Helicase A/Dhx9 in 10 µg of HeLa nuclear extract.

Target description

~140 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Shen et al.
Journal of cellular and molecular medicine, 25(1), 284-296 (2020-11-26)
Results from various studies reveal that the role of G protein-coupled oestrogen receptor (GPER) is cancer-context dependent, and the function of GPER in non-small-cell lung cancer (NSCLC) is still unclear. The present study demonstrated that neoplasm lung tissues expressed higher

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service