Skip to Content
Merck
All Photos(2)

Key Documents

MABE355

Sigma-Aldrich

Anti-ADAR2 Antibody, clone 2D1.1

clone 2D1.1, 1 mg/mL, from mouse

Synonym(s):

Nuclear pore complex protein Nup98-Nup96, 98 kDa nucleoporin, Nucleoporin Nup98, Nup98

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$657.00

$657.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$657.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$657.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D1.1, monoclonal

species reactivity

human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NUP98(4928)

General description

ADAR, also known as Nuclear pore protein 96 (NUP96) named after its molecular weight, or often in the literature as its yeast name, Nup145C, is one of them and NUP96 is made from the autocatalytic cleavage of a larger precursor protein named the Nuclear Pore Complex protein Nup98-Nup96, which also happens to be the source of NUP98 another important nuclear pore protein. NUC96 is found on both the nuclear and cytoplasmic sides of the NPC, whereas NUP98 apparently shuttles between the cytoplasmic side and the nucleoplasmic side of the NPC. Nup96 is part of a NP heptameric complex in the nuclear pore complex. Nup96 complexes with Sec13 in the middle of the heptamer complex. The function of the heptamer is to coat the curvature of the nuclear pore complex between the inner and outer nuclear membranes. Nup96 is predicted to be an alpha helical solenoid. The interaction between Nup96 and Sec13 is at the point of curvature in the heptameric complex. Interestingly, NUP96 and NUP98 also have a crucial role in the function of the immune system and are up regulated by interferons in response to viral infections. A heterozygous mutation in NUP96 has been shown to affect the expression of interferon-regulated genes and to disrupt the normal function of macrophages, and B and T lymphocytes, with no detectable gross pathological abnormalities in other organs. Moreover, the role of Nup96 in the immune response seems to be evolutionarily conserved, as NUP96 in Arabidopsis thaliana has also been shown to be required for the basal defense and resistance responses mediated by plant immune sensors.

Immunogen

GST-tagged recombinant protein corresponding to human ADAR2.

Application

Anti-ADAR2 Antibody, clone 2D1.1 is an antibody against ADAR2 for use in western blotting & ICC.
Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected ADAR2 in HeLa cells.

Alexa Fluor is a registered trademark of Life Technologies.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ADAR2 in 10 µg of HeLa cell lysate.

Target description

~98 kDa observed. Uniprot describes 6 isoforms due to alternative splicing at 198 kDa, 187 kDa, 98 kDa, 96 kDa, 196 kDa, and 186 kDa Uniprot also describes that the Nuclear pore complex protein Nup98-Nup96 is cleaved into a 98 kDa and a 96 kDa form. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service