Skip to Content
Merck
All Photos(2)

Key Documents

MABE224

Sigma-Aldrich

Anti-ZFX Antibody, clone 3E8.1

clone 3E8.1, from mouse

Synonym(s):

Zinc finger X-chromosomal protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$511.00

$511.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$511.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$511.00


Estimated to ship on30 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E8.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZFX(7543)

General description

ZFX, also known as Zinc finger X-chromosomal protein, and encoded by the gene name ZFX, is probable transcriptional activator. This gene on the X chromosome is structurally similar to a related gene on the Y chromosome. It encodes a member of the krueppel C2H2-type zinc-finger protein family. The full-length protein contains an acidic transcriptional activation domain (AD), a nuclear localization sequence (NLS) and a DNA binding domain (DBD) consisting of 13 C2H2-type zinc fingers. Studies in mouse embryonic and adult hematopoietic stem cells showed that this gene was required as a transcriptional regulator for self-renewal of both stem cell types, but it was dispensable for growth and differentiation of their progeny. Multiple alternatively spliced transcript variants encoding different isoforms have been identified, but the full-length nature of some variants has not been determined.

Immunogen

GST-tagged recombinant protein corresponding to human ZFX.

Application

Immunocytochemistry Analysis: 2.0 µg/mL from a representative lot detected ZFX HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-ZFX antibody is validated for use in WB, IC for the detection of ZFX.

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ZFX in 200 µg of HeLa nuclear extract.

Target description

~130, ~90, ~59 kDa observed. Three bands (~130 kDa, ~91 kDa and ~59 kDa) corresponding to ZFX were detected. The ~130 kDa band is nuclear, while the ~91 kDa band was found in the cytoplasmic (Zhu, Z., et al. (2013). J Neurooncol. 112:17-25).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service