Skip to Content
Merck
All Photos(1)

Key Documents

CLS430282

Corning® Easy-Grip round, plastic, storage bottles

round clear polystyrene, sterile, bottle capacity 500 mL, cap (plug seal), case of 24

Synonym(s):

storage bottle

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$28.70
24 EA
$114.00

$28.70


Check Cart for Availability


Select a Size

Change View
2 EA
$28.70
24 EA
$114.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

$28.70


Check Cart for Availability

material

cap (plug seal)
clear polystyrene bottle
cylindrical bottle
orange high-density polyethylene cap (plug seal)
round clear polystyrene

sterility

sterile
sterile

feature

cap
graduated
graduations
neck thread wide mouth

packaging

case of 24

manufacturer/tradename

Corning 430282

bottle capacity

500 mL

bottle neck diam.

~42 mm

neck diam.

45 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® Easy-Grip disposable storage bottles are ideal for storing media, buffers, and other aqueous solutions.
  • Features easy grip sides for convenient handling.
  • Plug seal caps (45 mm) provides an airtight seal and help minimize the risk of contamination.
  • Certified non pyrogenic.

Application

Corning®Easy-Grip round, plastic, storage bottles are ideal for storage of media, buffers, and other aqueous solutions.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service