Skip to Content
Merck
All Photos(1)

Key Documents

MABE1828

Sigma-Aldrich

Anti-beta-Catenin Antibody, clone mAb-Cat1

clone mAb-Cat1, from rat

Synonym(s):

beta-Catenin 1, Catenin beta-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$214.00
100 μL
$566.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$214.00
100 μL
$566.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mAb-Cat1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), bovine (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

ChIP: suitable
dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CTNNB1(1499)
mouse ... Ctnnb1(12387)

General description

Catenin beta-1 (UniProt: P35222; also known as beta-Catenin) is encoded by the CTNNB1 (also known as CTNNB, OK/SW-cl.35, PRO2286) gene (Gene ID: 1499) in human. Catenin beta-1 is a key downstream component of the canonical Wnt signaling pathway that participates in intercellular adhesion and Wnt-mediated transcriptional activation. In the absence of Wnt, it forms a complex with AXIN1, AXIN2, APC, CSNK1A1, and GSK-3 beta that promotes phosphorylation on N-terminal serine and threonine residues that leads to its subsequent degradation by the proteasome. In the presence of Wnt ligand, it is not ubiquitinated and accumulates in the nucleus, where it acts as a coactivator for transcription factors of the TCF/LEF family, which leads to activation of Wnt responsive genes. O-glycosylation at serine 23 is reported to decreases its nuclear localization and transcriptional activity. Wnt/beta-catenin signaling is shown to be essential for embryonic stem cell (ESC) pluripotency and differentiation. In ESC beta-catenin is trimethylated on Lysine 49 by Ezh2 methyltransferase and this trimethylated form represses neuronal differentiation genes Sox1 and Sox3. Mutations in CTNNB1 gene are shown to cause colorectal cancers, medulloblastoma, and ovarian cancers. (Ref.: Hoffmeyer, K., et al. (2017). Cell Reports 18(12); 2815-2824.

Specificity

This rat monoclonal antibody detects beta-catenin 1 unmodified and methylated or acetylated on lysine 49. It targets an epitope with in 19 amino acids from the N-terminal region.

Immunogen

BSA-conjugated linear peptide corresponding to 19 amino acids from the N-terminal region of human beta-catenin 1.

Application

Anti-beta-Catenin, clone mAb-Cat1, Cat. No. MABE1828, is a highly specific rat monoclonal antibody that targets beta-catenin 1 and has been tested for use in Chromatin Immunoprecipitation (ChIP), Dot Blot, Immunoprecipitation, and Western Blottiing.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected beta-Catenin in Western Blotting applications (Hoffmeyer, K., et. al. (2017). Cell Rep. 18(12):2815-2824).

Immunoprecipitation Analysis: A representative lot immunoprecipitated beta-Catenin in Immunoprecipitation applications (Hoffmeyer, K., et. al. (2017). Cell Rep. 18(12):2815-2824).

Dot Blot Analysis: A representative lot detected beta-Catenin in Dot Blot applications (Hoffmeyer, K., et. al. (2017). Cell Rep. 18(12):2815-2824).

Chromatin Immunoprecipitation Analysis (ChIP): A representative lot immunoprecipitated beta-Catenin in Chromatin Immunoprecipitation applications (Hoffmeyer, K., et. al. (2017). Cell Rep. 18(12):2815-2824).

Quality

Evaluated by Western Blotting in SW 480 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected beta-Catenin in SW 480 cell lysate.

Target description

~100 kDa observed; 85.48 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service