Skip to Content
Merck
All Photos(2)

Key Documents

MABE1791M

Sigma-Aldrich

Anti-KAT2A (GCN5) Antibody, clone 2GC-2C11

clone 2GC-2C11, from mouse

Synonym(s):

Histone acetyltransferase KAT2A, EC: 2.3.1.48, General control of amino acid synthesis protein 5-like 2, Histone acetyltransferase GCN5, HsGCN5, Lysine acetyltransferase 2A, STAF97

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$214.00
100 μG
$512.00

$214.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$214.00
100 μG
$512.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2GC-2C11, monoclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... KAT2A(2648)

General description

Histone acetyltransferase KAT2A (UniProt: Q92830; also known as EC: 2.3.1.48, General control of amino acid synthesis protein 5-like 2, Histone acetyltransferase GCN5, HsGCN5, Lysine acetyltransferase 2A, STAF97) is encoded by the KAT2A (also known as GCN5, GCN5L2, HGCN5) gene (Gene ID: 2648) in human. Histone acetyltransferase KAT2A belongs to the acetyltransferase family and is expressed in all tissues tested. However, its expression is highest in ovaries. It has three acetyl-CoA-binding regions (aa 579-581; 586-592; 617-620) and one bromodomain. It functions as a histone acetyltransferase (HAT) to promote transcriptional activation. Acetylation of histones gives a specific tag for epigenetic transcription activation. It displays significant histone acetyltransferase activity with core histones, but not with nucleosome core particles. KAT2A is also shown to acetylate non-histone proteins, such as CCAAT/enhancer-binding protein beta (CEBPB). In case of HIV-1 infection, histone acetyltransferase KAT2A is recruited by the viral protein Tat and it can regulates Tat′s transactivating activity and may help in inducing chromatin remodeling of proviral genes. Two isoforms of histone acetyltransferase KAT2A have been reported that are produced by alternative splicing. Isoform 2 lacks the first 410 amino acids.


Specificity

Clone 2GC-2C11 detects Histone acetyltransferase KAT2A in human and murine cells. It targets an epitope within 15 amino acids from the N-terminal region.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human Histone acetyltransferase KAT2A.

Application

Anti-KAT2A (GCN5), clone 2GC-2C11, Cat. No. MABE1791, is a highly specific mouse monoclonal antibody that targets Histone acetyltransferase KAT2A and has been tested in Western Blotting and Immunoprecipitation.
Western Blotting Analysis: 2 µg/mL from a representative lot detected KAT2A (GCN5) in 10 µg of NIH/3T3 cell lysate.

Immunoprecipitation Analysis: A representative lot detected KAT2A (GCN5) in Immunoprecipitation applications (Brand, M., et. al. (2001). EMBO J. 20(12):3187-96).

Western Blotting Analysis: A representative lot detected KAT2A (GCN5) in Western Blotting applications (Riss, A., et. al. (2015). J Biol Chem. 290(48):28997-9009; Helmlinger, D., et. al. (2006). PLoS Biol. 4(3):e67; Brand, M., et. al. (2001). EMBO J. 20(12):3187-96).

Quality

Evaluated by Western Blotting in HeLa nuclear extracts.

Western Blotting Analysis: 2 µg/mL of this antibody detected KAT2A (GCN5) in 10 µg of HeLa cells nuclear extract.

Target description

~94 kDa observed; 93.93 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service